Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Corning (85)
Remel (74)
Product Type:
Agar (100)
Broths (53)
Availability:
Capacity (g):
Capacity (mL):
0 - 20 (2)
Material:
Glass (20)
Capacity (Liters):
0 to 1 (17)
Sterile:
Yes (13)
Max RCF (x g):
25,314 (12)
17,000 (2)
Max rpm:
15,200 (12)
13,300 (1)
13300 (1)
Well shape:
Round (7)
Flat (6)
Qty/pk:
100 (6)
Contained Volume:
1 µL (4)
10 µL (3)
1µL (1)
No. of wells:
6 (3)
12 (2)
24 (2)
96 (2)
48 (1)
4 (1)
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Life Science > Microbiology

Microbiology

Microbiology

Cole-Parmer has been providing laboratory solutions to all industries for over 50 years. Known for our exceptional Technical Assistance and Customer Service, we have been providing the right products for every unique situation.

Besides our well known line of Masterflex pumps and tubing, Cole-Parmer also offers a full line of microbiology products for the Life Scientist.

Products offered for microbiology applications include culture media, innoculating loops, petri dishes and contact plates, bacteria test kits, and air samplers. Microbiology refers to the study of microorganisms, or those organisms that are too small to view with the human eye.

Industries utilizing Microbiology Products

  • Pharmaceutical
  • Food
  • Industrial
  • Chemical
  • Water and Wastewater

 Product Detail
Sort
 Availability
 Price
EW-06139-00 Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/box

Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/box

Product Type Culture Dishes/Plates
EW-06139-00
In stock.
$135.00 USD/BOX of 500

Qty:

EW-39152-80 MAS-100Eco air sampler

MAS-100Eco air sampler

Product Type Air Samplers,Brand MAS
EW-39152-80
Usually ships in 14 days.
$4,770.00 USD/EACH

Qty:

EW-17710-46 Save over $5200 with this centrifuge bundle!
EW-17710-46
Usually ships in 6 days.
$13,455.00 USD/EACH

Qty:

EW-36150-00 Dilution bottle, Butterfield's phosphate buffer, 99 mL fill capacity

Dilution bottle, Butterfield's phosphate buffer, 99 mL fill capacity

Product Type Dilution bottles,Material Polypropylene,Capacity Liters 0.145
EW-36150-00
Usually ships in 5 days.
$94.00 USD/PACKAGE

Qty:

EW-33687-02 Microbial air sampler, digital, stainless steel head, 115 VAC

Microbial air sampler, digital, stainless steel head, 115 VAC

Product Type Air Samplers,Power VAC 115,Brand SAS
EW-33687-02
Usually ships in 13 days.
$6,436.00 USD/EACH

Qty:

EW-14207-70 Representative image only.  Check individual details.

Thermo Scientific Contact plate sterile tryptic soy agar with lecithin, polysorbate 80, 10/PK

Product Type Prepared Media Plates,Description Irradiated. For cultivation of bacteria, yeast, and fungi.,Media Contact plate sterile tryptic soy agar with lecithin, polysorbate 80,Qty/pk 10
EW-14207-70
Usually ships in 4 days.
$42.75 USD/PACKAGE

Qty:

EW-14040-30 VERIcleen Rapid Surface Hygiene Kit

VERIcleen Rapid Surface Hygiene Kit

Product Type Bacterial Test Kit,Brand VERIclean
EW-14040-30
Usually ships in 4 days.
$100.00 USD/EACH

Qty:

Representative photo only Prepared agar plate PDA potato dextrose agar 10 pk

Prepared agar plate, PDA (potato dextrose agar),10/pk

Product Type Agar,Description PDA (Potato Dextrose Agar) for cultivation of yeasts and mold and for the inducement of sporulation.,Media Name Media P-R
EW-14201-52
Usually ships in 8 days.
$16.50 USD/PACKAGE

Qty:

Representative photo only Sterile Petri Dishes 60 mm dia x 15 mm H 500 box

Sterile Petri Dishes; 60 mm dia x 15 mm H; 500/box

Product Type Culture Dishes/Plates
EW-06139-01
In stock.
$140.00 USD/BOX of 500

Qty:

Representative photo only Pyrex Brand 3160 petri dish 150 x 20 mm pack of 12

Pyrex Brand 3160 petri dish; 150 x 20 mm, pack of 12

Product Type Petri Dishes,Material Glass,Brand Pyrex
EW-34551-06
In stock.
$241.00 USD/PACKAGE of 12

Qty:

Representative photo only Thermo Scientific Nunc Lab Tek Sterile Petri Dishes PS Standard 100mm x 15 mm 320 case

Thermo Scientific Nunc Lab-Tek Sterile Petri Dishes, PS, Standard, 100mm x 15 mm; 320/case

Product Type Petri Dishes,Material Disposable polystyrene,Description Standard
EW-14007-01
In stock.
$165.00 USD/CASE

Qty:

Representative photo only Prepared agar plate TSA 10 pk

Prepared agar plate, TSA,10/pk

Product Type Agar,Description TSA, general growth medium for the isolation and cultivation of microorganisms.,Media Name Media P-R
EW-14201-42
Usually ships in 12 days.
$13.00 USD/PACKAGE

Qty:

Representative photo only Thermo Scientific Nunc Thermo Scientific Nunclon 48 well MultiDish with lid PS sterile

Thermo Scientific Nunc Thermo Scientific Nunclon 48-well MultiDish with lid, PS, sterile

Product Type Culture Dishes/Plates,No. of wells 48,Brand Thermo Scientific Nunc
EW-01930-24
In stock.
$330.00 USD/CASE of 75

Qty:

EW-14201-40 Prepared agar plate, 5% blood agar,10/pk

Prepared agar plate, 5% blood agar,10/pk

Product Type Agar,Description Blood Agar, 5%, general purpose growth media for the isolation, cultivation, and differentiation of a wife variety of microorgansim.,Media Name Media P-R
EW-14201-40
Usually ships in 5 days.
$13.00 USD/PACKAGE

Qty:

EW-14207-00 Representative image only.  Check individual details.

Thermo Scientific Tryptic soy agar (TSA), soybean casein digest, 10/pk

Product Type Prepared Media Plates,Description General-purpose medium for cultivation and isolation of microorganisms. Industrial reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW,Media Tryptic soy agar (TSA), soybean casein digest,Qty/pk 10
EW-14207-00
Usually ships in 7 days.
$8.05 USD/PACKAGE

Qty:

617 results (showing 1 - 15 )
Per Page 
Home  >  Life Science > Microbiology

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title