log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Microfluidic Pumps
Microfluidic Pumps
Microfluidic Pumps
A microfluidic device was once only the domain of inkjet printers and similarly-styled office equipment. But flash forward to today and you will see a microfluidic device employed in biotechnology, pharmaceutical and life sciences industry. A microfluidic device controls the flow of liquids and gases. This type if pump technology is a vital stopgap in many applications. Fortunately there are a few different pump technologies that are capable of reaching the microliter flow range. Most prevalent would be the syringe pump technology. Other micro pumps include peristaltic or diaphragm pumps. Starting with the syringe pumps, one will find a wide range of options. Many of these are capable of achieving single digit picoliter per minute flow rates – which is ideal for microfluidic devices. The syringe pumps also offer a more uniform flow with less pulsation in the delivered fluid. Another possible technology for micro pump applications would be the small self-priming micro diaphragm pumps that are able to dispense down to 20 microliters per stroke. Still another option is the new Cole-Parmer micro peristaltic pumps. These pumps also offer a graphical user interface to help control the pump and ensure accuracy and repeatability to such low flow rates. If you need to know more about purchasing the right microfluidic device contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
EW-74900-05 Infusion pump
EW-74900-05
In stock.
$1,542.00 USD/EACH
Qty:
EW-74900-00 Infusion pump
EW-74900-00
In stock.
$1,371.00 USD/EACH
Qty:
EW-78023-32 Representative image only.  Check individual details.
Ismatec Low-Speed, Planetary Gear-Driven, Digital Pump; 16 channel
Product Type Planetary Gear-Driven,Channels 16,Rollers 8
EW-78023-32
Call 1-888-358-4705 for availability.
$3,809.00 USD/EACH
Qty:
EW-73150-00 Piston Metering Pump, 3/32" stainless head, 0.002 to 2.5 mL/min range, 6000 psi
Piston Metering Pump, 3/32" stainless head, 0.002 to 2.5 mL/min range, 6000 psi
Product Type Piston Metering Pumps,Max pressure 6000 psi,CE Compliance Yes
EW-73150-00
Usually ships in 12 days.
$2,441.00 USD/EACH
Qty:
Representative photo only Piston Metering Pump 1 8 stainless head 0 01 to 10 mL min range 6000 psi Damper
Piston Metering Pump, 1/8" stainless head, 0.01 to 10 mL/min range, 6000 psi, Damper
Product Type Piston Metering Pumps,Max pressure 6000 psi,CE Compliance Yes
EW-73150-42
Usually ships in 11 days.
$3,247.00 USD/EACH
Qty:
Representative photo only Piston Metering Pump 1 8 stainless head 0 01 to 10 mL min range 6000 psi
Piston Metering Pump, 1/8" stainless head, 0.01 to 10 mL/min range, 6000 psi
Product Type Piston Metering Pumps,Max pressure 6000 psi,CE Compliance Yes
EW-73150-40
Call 1-888-358-4705 for availability.
$2,337.00 USD/EACH
Qty:
EW-78023-02 Ismatec<small><sup>&reg;</sup></small> Low-Speed, Planetary Gear-Driven, Digital Pump; 4 channel
Ismatec Low-Speed, Planetary Gear-Driven, Digital Pump; 4 channel
Product Type Planetary Gear-Driven,Channels 4,Rollers 8
EW-78023-02
Call 1-888-358-4705 for availability.
$2,957.00 USD/EACH
Qty:
EW-78023-22 Representative image only.  Check individual details.
Ismatec Low-Speed, Planetary Gear-Driven, Digital Pump; 12 channel
Product Type Planetary Gear-Driven,Channels 12,Rollers 8
EW-78023-22
Call 1-888-358-4705 for availability.
$3,564.00 USD/EACH
Qty:
EW-78023-42 Representative image only.  Check individual details.
Ismatec Low-Speed, Planetary Gear-Driven, Digital Pump; 24 channel
Product Type Planetary Gear-Driven,Channels 24,Rollers 8
EW-78023-42
Usually ships in 15 days.
$4,165.00 USD/EACH
Qty:
Representative photo only Piston Metering Pump 1 8 stainless head 0 003 to 5 mL min range 6000 psi Damper
Piston Metering Pump, 1/8" stainless head, 0.003 to 5 mL/min range, 6000 psi, Damper
Product Type Piston Metering Pumps,Max pressure 6000 psi,CE Compliance Yes
EW-73150-12
Call 1-888-358-4705 for availability.
$3,247.00 USD/EACH
Qty:
EW-73160-20 Model 73160-00 pictured
Low Flow Peristaltic Pump, Variable Speed, 8 - 22 RPM, 12 VDC, Manual Control
Product Type Peristaltic Metering Pumps,Max pressure 10 psi,Min flow rate mL/min 0.14
EW-73160-20
Call 1-888-358-4705 for availability.
$203.00 USD/EACH
Qty:
Representative photo only Piston Metering Pump 3 32 stainless head 0 002 to 2 5 mL min range 6000 psi Damper
Piston Metering Pump, 3/32" stainless head, 0.002 to 2.5 mL/min range, 6000 psi, Damper
Product Type Piston Metering Pumps,Max pressure 6000 psi,CE Compliance Yes
EW-73150-02
Call 1-888-358-4705 for availability.
$3,350.00 USD/EACH
Qty:
EW-73100-50 Microannular Gear Pump, 5microliter to 255ml/min, 1160 PSI with digital controller
Microannular Gear Pump, 5microliter to 255ml/min, 1160 PSI with digital controller
Product Type Gear Pump,Min flow rate mL/min 0.048,Max flow rate mL/min 288
EW-73100-50
Call 1-888-358-4705 for availability.
$14,220.00 USD/EACH
Qty:
Representative photo only Piston Metering Pump 1 4 stainless head 0 01 to 20 mL min range 3000 psi
Piston Metering Pump, 1/4" stainless head, 0.01 to 20 mL/min range, 3000 psi
Product Type Piston Metering Pumps,Max pressure 3000 psi,CE Compliance Yes
EW-73150-20
Usually ships in 13 days.
$2,683.00 USD/EACH
Qty:
Representative photo only Cole Parmer Inert Solenoid Self Priming Micro Pumps 50 microL 24 VDC PPS EPDM
Cole-Parmer Inert Solenoid Self Priming Micro Pumps, 50 microL, 24 VDC, PPS, EPDM
Product Type Diaphragm Pumps,Max pressure 5 psi,Max temperature º C 70
EW-73120-24
Call 1-888-358-4705 for availability.
$238.00 USD/EACH
Qty:
73 results (showing 1 - 15 )
Per Page 
Home  >  Pumps > Microfluidic Pumps
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title