Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Microplate Readers

Microplate Readers

Microplate Readers
Choose from our comprehensive line of Microplate Readers and other Immunology products.
 Product Detail
Sort
 Availability
 Price
Microplate reader 340 to 750 nm 6 12 24 48 96 well (Representative photo only)

Microplate reader, 340 to 750 nm, 6, 12, 24, 48, 96 well

Product Type Microplate reader,Brand BioTek
EW-13055-54
Available for order
or call 1-888-358-4705 to order
$5,805.00 USD/EACH

Qty:

EW-13055-50 Microplate reader, 400 to 750 nm, 6, 12, 24, 48, 96 well

Microplate reader, 400 to 750 nm, 6, 12, 24, 48, 96 well

Product Type Microplate reader,Brand BioTek
EW-13055-50
Usually ships in 14 days.
$5,500.00 USD/EACH

Qty:

Microplate reader 400 to 750 nm 6 12 24 48 72 96 Terasaki 96 384 well (Representative photo only)
EW-13055-52
Available for order
or call 1-888-358-4705 to order
$6,365.00 USD/EACH

Qty:

3 results (showing 1 - 3 )
Per Page 
Home  >  Microplate Readers