Home  >  Pressure & Vacuum Instruments > Pressure Regulators for Gases > Mini High-Purity Pressure Regulators
Mini High-Purity Pressure Regulators
Mini High-Purity Pressure Regulators
  • Compact, single stage regulators ideally suited for lecture bottles and other small cylinders
  • Sensitive diaphragm provides accurate pressure settings
  • 1 ½“ Gauge permits easy reading
  • Large adjusting knob provides excellent sensitivity over a wide range of settings
For use with non- or mildly- corrosive gasses, these regulators are equipped with a CGA 180 swivel connection for easy attachment, and a built- in, non-lubricated needle valve designed to regulate outlet flow. Choose a brass model for non-analytical, non-corrosive service, an aluminum model for mildly corrosive service, or a stainless steel model for corrosive service.
 Product Detail
Sort
 Availability
 Price
Representative photo only Mini High Purity Pressure Regulators brass
Mini High-Purity Pressure Regulators, brass
Product Type High Purity Gas Regulators,Max psi inlet 3000,Capacity SCFH 1050
EW-98202-38
Usually ships in 7 days.
$309.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Pressure & Vacuum Instruments > Pressure Regulators for Gases > Mini High-Purity Pressure Regulators
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title