Home  >  Flow, Level & Valves > Valves > Check Valves > Miniature Check Valves
Miniature Check Valves
Miniature Check Valves
  • Use with liquids and gases for pressure or vacuum applications
  • Breaking pressure is 0.18 psi, closing pressure is 0.014 psi
  • Install in any position
Note: Valves require 1 psi back pressure for a tight seal.
 Product Detail
Sort
 Availability
 Price
EW-98553-10 Representative Photo Only.  PP Miniature Check Valve, 1/8" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm
PP Miniature Check Valve, 1/8" hose barb, Viton diaphragm
Product Type Check Valves (manual),Cv 0.11,Material polypropylene
EW-98553-10
In stock.
$9.75 USD/EACH
Qty:
EW-98553-12 Representative Photo Only.  PP Miniature Check Valve, 1/4" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm
PP Miniature Check Valve, 1/4" hose barb, Viton diaphragm
Product Type Check Valves (manual),Cv 0.12,Material polypropylene
EW-98553-12
In stock.
$5.75 USD/EACH
Qty:
EW-98553-02 Representative Photo Only.  Nylon Miniature Check Valve, 1/4" hose barb, fluorosilicone diaphragm
Nylon Miniature Check Valve, 1/4" hose barb, fluorosilicone diaphragm
Product Type Check Valves (manual),Cv 0.13,Material nylon
EW-98553-02
In stock.
$4.50 USD/EACH
Qty:
EW-98553-22 Representative Photo Only.  PVDF Miniature Check Valve, 1/4" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm
PVDF Miniature Check Valve, 1/4" hose barb, Viton diaphragm
Product Type Check Valves (manual),Cv 0.12,Material PVDF
EW-98553-22
In stock.
$8.75 USD/EACH
Qty:
EW-98553-01 Representative Photo Only.  Nylon Miniature Check Valve, 3/16" hose barb, fluorosilicone diaphragm
Nylon Miniature Check Valve, 3/16" hose barb, fluorosilicone diaphragm
Product Type Check Valves (manual),Cv 0.13,Material nylon
EW-98553-01
In stock.
$4.50 USD/EACH
Qty:
EW-98553-00 Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm
Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm
Product Type Check Valves (manual),Cv 0.12,Material nylon
EW-98553-00
In stock.
$4.50 USD/EACH
Qty:
EW-98553-11 Representative Photo Only.  PP Miniature Check Valve, 3/16" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm
PP Miniature Check Valve, 3/16" hose barb, Viton diaphragm
Product Type Check Valves (manual),Cv 0.12,Material polypropylene
EW-98553-11
In stock.
$5.75 USD/EACH
Qty:
EW-98553-34 Representative Photo Only.  PVDF Miniature Check Valve, 3/8" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm
PVDF Miniature Check Valve, 3/8" hose barb, Viton diaphragm
Product Type Check Valves (manual),Cv 0.12,Material PVDF
EW-98553-34
In stock.
$9.75 USD/EACH
Qty:
EW-98553-13 Representative Photo Only.  PP Miniature Check Valve, 5/16" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm
PP Miniature Check Valve, 5/16" hose barb, Viton diaphragm
Product Type Check Valves (manual),Cv 0.13,Material polypropylene
EW-98553-13
In stock.
$8.50 USD/EACH
Qty:
EW-98553-21 Representative Photo Only.  PVDF Miniature Check Valve, 3/16" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm
PVDF Miniature Check Valve, 3/16" hose barb, Viton diaphragm
Product Type Check Valves (manual),Cv 0.12,Material PVDF
EW-98553-21
In stock.
$9.00 USD/EACH
Qty:
10 results (showing 1 - 10 )
Per Page 
Home  >  Flow, Level & Valves > Valves > Check Valves > Miniature Check Valves
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title