log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Balances & Scales > Moisture Balances
Moisture Balances
Moisture Balances
A moisture analyzer is perfect for measuring moisture in most products including grains, pharmaceuticals, soils, sludge and chemicals. The amount of moisture in a product can greatly affect perceived quality. You use a moisture analyzer to measure the original weight of a product before the drying process. The machine takes note of that weight and then after the drying process it notates the decrease in moisture and shows the weight change on the moisture analyzer display. Heating lamps are sometimes used to speed up the drying process. Ceramic heating elements are designed to rapidly attain the final temperature to ensure a uniform distribution of heat over the entire surface, thus eliminating localized burning of the sample. Halogen lamp heating elements are suited for applications that require accelerated temperature regulation during sample drying. However halogen lamps heat and cool quickly sometimes causing the sample to scorch. A moisture analyzer can also use quartz heating elements, which looks similar to the heating element in your oven at home. Heat is transferred from a coil wire to a material which distributes the heat providing consistent heating. There are many other factors in selecting a moisture analyzer but these are essential.
 Product Detail
Sort
 Availability
 Price
EW-01002-81 Ohaus MB Moisture Determination Balance, Mb23 110 G X 0.01 G 120VAC
Ohaus MB Moisture Determination Balance, Mb23 110 G X 0.01 G 120VAC
Product Type Moisture Balances,Calibration External,Capacity g 110
EW-01002-81
In stock.
$2,595.00 USD/EACH
Qty:
EW-11213-98 Sartorius MA Moisture Determination Balance, Ceramic Infrared heater, 35g x 0.001%, 115VAC
Sartorius MA Moisture Determination Balance, Ceramic Infrared heater, 35g x 0.001%, 115VAC
Product Type Moisture Balances,Calibration External,Capacity g 35
EW-11213-98
Usually ships in 14 days.
$3,160.00 USD/EACH
Qty:
EW-01002-84 Ohaus MB Moisture Determination Balance, Mb45 45 G  X  0.001 G 120VAC
Ohaus MB Moisture Determination Balance, Mb45 45 gX 0.001 G 120VAC
Product Type Moisture Balances,Calibration External,Capacity g 45
EW-01002-84
Usually ships in 12 days.
$4,195.00 USD/EACH
Qty:
EW-11334-48 METTLER TOLEDO Moisture Determination Balance, 54 g x 0.01%
Mettler Toledo Moisture Determination Balance, 54 g x 0.01%
Product Type Moisture Balances,Calibration External,Capacity g 54
EW-11334-48
Usually ships in 7 days.
$4,347.00 USD/EACH
Qty:
EW-01041-30 A&D Moisture Determination Balance, MF-50, 51 g x 0.002 g, 115 VAC.  Representative Photo Only
A&D Moisture Determination Balance, MF-50, 51 g x 0.002 g, 115 VAC
Product Type Moisture Balances,Calibration External,Capacity g 51
EW-01041-30
Usually ships in 6 days.
$2,725.00 USD/EACH
Qty:
EW-11702-62 Adam PMB Moisture Determination Balance, 200g X 01g/0.05%, 110 VAC.  Representative Photo Only
Adam PMB Moisture Determination Balance, 200g X 01g/0.05%, 110 VAC
Product Type Moisture Balances,Calibration External,Capacity g 200
EW-11702-62
Usually ships in 10 days.
$1,795.00 USD/EACH
Qty:
EW-11124-06 Cole-Parmer® Symmetry® MB Moisture Determination Balance, 40g x 0.001g/0.01% Moisture, 115V
Cole-Parmer Symmetry MB Moisture Determination Balance, 40g x 0.001g/0.01% Moisture, 115V
Product Type Moisture Balances,Calibration External,Capacity g 40
EW-11124-06
Usually ships in 6 days or sooner.
$1,895.00 USD/EACH
Qty:
See all
EW-01041-40 A&D Moisture Determination Balance, MX-50, 51 g x 0.001 g, 115 VAC.  Representative Photo Only
A&D Moisture Determination Balance, MX-50, 51 g x 0.001 g, 115 VAC
Product Type Moisture Balances,Calibration External,Capacity g 51
EW-01041-40
Usually ships in 4 days.
$3,395.00 USD/EACH
Qty:
EW-11334-47 METTLER TOLEDO Moisture Determination Balance, 35 g x 0.01%
Mettler Toledo Moisture Determination Balance, 35 g x 0.01%
Product Type Moisture Balances,Calibration External,Capacity g 35
EW-11334-47
Usually ships in 7 days.
$3,347.00 USD/EACH
Qty:
EW-01002-83 Ohaus MB Standard Moisture Determination Balance, Mb35 35 G X 0.002 G 120VAC
Ohaus MB Standard Moisture Determination Balance, Mb35 35 G X 0.002 G 120VAC
Product Type Moisture Balances,Calibration External,Capacity g 35
EW-01002-83
In stock.
$3,675.00 USD/EACH
Qty:
EW-01041-25 A&D Moisture Determination Balance, ML-50, 51 g x 0.005 g, 115 VAC.  Representative Photo Only
A&D Moisture Determination Balance, ML-50, 51 g x 0.005 g, 115 VAC
Product Type Moisture Balances,Calibration External,Capacity g 51
EW-01041-25
Usually ships in 13 days.
$2,295.00 USD/EACH
Qty:
EW-11702-60 Adam PMB Moisture Determination Balance, 50g X 1mg/0.01%, 110 VAC.  Representative Photo Only
Adam PMB Moisture Determination Balance, 50g X 1mg/0.01%, 110 VAC
Product Type Moisture Balances,Calibration External,Capacity g 50
EW-11702-60
Usually ships in 10 days.
$2,195.00 USD/EACH
Qty:
EW-11702-61 Adam PMB Moisture Determination Balance, 50g X 1mg/0.01%, 220 VAC. Representative Photo Only
Adam PMB Moisture Determination Balance, 50g X 1mg/0.01%, 220 VAC
Product Type Moisture Balances,Calibration External,Capacity g 50
EW-11702-61
Usually ships in 10 days.
$2,195.00 USD/EACH
Qty:
EW-11124-07 Cole-Parmer® Symmetry® MB Moisture Determination Balance, 40g x 0.001g/0.01% Moisture, 220V.  Representative Photo Only
Cole-Parmer Symmetry MB Moisture Determination Balance, 40g x 0.001g/0.01% Moisture, 220V
Product Type Moisture Balances,Calibration External,Capacity g 40
EW-11124-07
Usually ships in 6 days.
$1,895.00 USD/EACH
Qty:
See all
EW-01041-45 A&D Moisture Determination Balance, MS-70, 71 g x 0.0001 g, 115 VAC.  Representative Photo Only
A&D Moisture Determination Balance, MS-70, 71 g x 0.0001 g, 115 VAC
Product Type Moisture Balances,Calibration External,Capacity g 71
EW-01041-45
Usually ships in 7 days.
$5,525.00 USD/EACH
Qty:
22 results (showing 1 - 15 )
Per Page 
Home  >  Balances & Scales > Moisture Balances
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title