Mops
Mops
Cleanroom mops are specialty tools and require special care when purchasing. Cleanroom mops are built for specific applications. Both the head and the handle are constructed as such. Cleanroom mops are used to clean controlled environment walls and floors. Mops have low contamination characteristics and should be laundered and packaged in order to avoid static, residue, and particulates. Handles can be steel coated with white polyester to ensure completely corrosion resistance. Choosing the right mop head is important as well. You want to make sure the head is compatible with disinfectants commonly used in controlled environments. Some lab managers like the cleanroom mops with a knitted head made from tubular polyester for its sturdiness. Others like mop heads that do not include adhesives to clean not only the floor but the walls as well. But there are also handheld roll mops which have adhesive coating to remove lint and dirt from plastic, metal and glass. A Cole-Parmer technical expert can help you choose cleanroom mops that fit with your controlled environment and your cleaning specifications.
 Product Detail
Sort
 Availability
 Price
EW-09411-60 Telescopic adhesive roll mop; 18" wide
Telescopic adhesive roll mop; 18" wide
Description Telescopic handle,Model 6-300
EW-09411-60
In stock.
$294.00 USD/EACH
Qty:
EW-09411-30 Contec EasySat Bucketless Floor Mop shown with Mop head (sold seperately).
Contec EasySat Bucketless Floor Mop
Product Type Mop,Description EasySat Bucketless Mop,Dimensions " L 57.5
EW-09411-30
Usually ships in 10 days.
$295.00 USD/EACH
Qty:
See all
Representative photo only Wall WipR Cleaning System Mop Cover Polyester Foam
Wall WipR Cleaning System, Mop Cover, Polyester Foam
Product Type Mops,Brand Contec,Model WWC18
EW-33675-61
In stock.
$94.00 USD/PACKAGE of 50
Qty:
EW-09411-35 Contec EasySat Bucketless Floor Mop shown with Mop Head (sold seperately).
Contec EasySat Textured Mop Head for Bucketless Floor Mop
Product Type Mop Head,Description Mop head, textured,Material Ultraclean polyester/ester foam
EW-09411-35
Usually ships in 10 days.
$322.00 USD/PACKAGE
Qty:
See all
EW-09411-37 Contec EasySat Bucketless Floor Mop shown with Mop Head (sold seperately).
Contec EasySat Sterile Textured Mop Head for Bucketless Floor Mop
Product Type Mop Head,Description Mop head, textured and sterile,Material Ultraclean polyester/ester foam
EW-09411-37
Usually ships in 10 days.
$478.00 USD/PACKAGE
Qty:
See all
Representative photo only Betamop II heads 6
Betamop II heads (6)
Product Type Mops,Description Betamop II heads (6),Brand TEXWIPE
EW-86518-22
Usually ships in 15 days.
$299.00 USD/CASE
Qty:
EW-09411-31 Contec EasySat Bucketless Floor Mop shown with Mop Head (sold seperately).
Contec EasySat Mop Head for Bucketless Floor Mop
Product Type Mop Head,Description Bucketless Floor Mop Head,Material Ultraclean polyester/ester foam
EW-09411-31
Usually ships in 10 days.
$310.00 USD/PACKAGE
Qty:
See all
Representative photo only Alphamop pads 6 and covers 150
Alphamop, pads (6) and covers (150)
Product Type Mops,Description Alphamop pads (6) and head covers (150),Brand TEXWIPE
EW-86518-02
Usually ships in 15 days.
$561.00 USD/CASE
Qty:
EW-09411-50 Handheld adhesive roll mop; 9" wide
Handheld adhesive roll mop; 9" wide
Description Hand-held,Model 6-301
EW-09411-50
In stock.
$62.00 USD/EACH
Qty:
Representative photo only Handheld adhesive roll mop 3 W roller
Handheld adhesive roll mop, 3"W roller
Description Telescopic handle,Model 6-303
EW-09411-40
Call 1-888-358-4705 for availability.
$56.00 USD/EACH
Qty:
EW-86518-20 Betamop II mop
Betamop II mop
Product Type Mops,Description Betamop II mop,Brand TEXWIPE
EW-86518-20
Usually ships in 15 days.
$98.00 USD/CASE
Qty:
Representative photo only Squeegee 24 W
Squeegee, 24"W
Product Type Mops,Color White,Model 9524
EW-86517-00
In stock.
$24.00 USD/EACH
Qty:
Representative photo only Adjustable handle 34 to 62 L
Adjustable handle, 34 to 62" L
Product Type Mops,Color White,Model 8800
EW-86517-10
In stock.
$21.50 USD/EACH
Qty:
Representative photo only Wall WipR Cleaning System Mop Head
Wall WipR Cleaning System, Mop Head
Product Type Mops,Brand Contec,Model WWH18
EW-33675-60
Usually ships in 8 days.
$26.00 USD/EACH
Qty:
EW-09411-36 Contec EasySat Bucketless Floor Mop shown with Mop Head (sold seperately).
Contec EasySat Sterile Mop Head for Bucketless Floor Mop
Product Type Mop Head,Description Mop head, sterile,Material Ultraclean polyester/ester foam
EW-09411-36
Usually ships in 10 days.
$452.00 USD/PACKAGE
Qty:
See all
18 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title