United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Radios
> Motorola Spirit Two-Way FM Radios
Motorola Spirit Two-Way FM Radios
Communicate with colleagues and work-groups in and around large buildings or industrial sites.
VHF radios operate in one of four frequencies. UHF radios operate in one of eight frequencies. Change frequencies and code settings with push-to-talk button. Access voice-assisted programming with monitor button.
VHF 1 Watt Radios are ideal for small to medium buildings. Transmit signal up to 150,000 sq ft and 8 floors in steel-and-concrete reinforced buildings; up to 5 miles outdoors in flat open areas with no obstructions.
UHF 2 Watt Radios are ideal for medium to large buildings. Transmit signal up to 250,000 sq ft and 20 floors in steel-and-concrete reinforced buildings; up to 5 miles outdoors in flat open areas with no obstructions.
All radios include an 8" antenna, belt clip, NiCd battery pack, battery charger adapter for 115 VAC power source, and an outlet jack to hook up speaker/microphone accessories (sold separately).
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Remote Speaker Microphone for Motorola Spirit radios
Description Shoulder mount microphone,Brand Motorola,Model 53862
EW-08983-38
Usually ships in 10 days.
$110.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Motorola Spirit Two-Way FM Radios
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Radios
> Motorola Spirit Two-Way FM Radios
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title