Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Motors
Motors
Motors
Faced motors are among the most common motors in installations. We offer a wide selection of faced motors, including an extensive selection of standard NEMA frame motors, to suit your application. IEC configurations are also available.
Motor Types
DC Motors are used in a variety of variable-speed applications.
Air Motors are inherently intrinsically safe due to lack of electrical power. Air motors, however, are not as efficient as electric motors. By controlling sourced air, motors offer variable speed control. Not all locations have source air available. If you need a compressed air source, see our ¡°Compressors¡± offerings.
AC Motors come in two versions¡ªsingle and three-phase powered. Single-phase power is available in residential and commercial locations, and is suitable for fixed-speed applications. Three-phase power enables you to get variable-speed control with an appropriate controller. We recommend that you consult a qualified electrician for any electrical installation.
Motor Enclosures
Open Drip-Proof (ODP) Motors have open enclosures, suitable for indoor use and clean atmospheres. The ventilator openings are constructed to prevent liquids and solids from entering the machine from an angle of 0 to 15¡ã from vertical.
Totally Enclosed, Fan-Cooled (TEFC) Motors are equipped with an external fan to circulate outside air to the motor.
Totally Enclosed, Nonvented) (TENV) Motors are designed without air openings. Although air does not freely move between the inside and outside of the motor, construction is not air- or liquid-tight.
Washdown (WDN) Enclosures are great for applications in environments where frequent cleaning or sanitizing is necessary. Provided in both white epoxy and stainless steel construction.
Explosion-Proof (XPRF) Motors are totally enclosed and designed to withstand an internal explosion of gas or vapor¡ªmotor frame will not burst or rupture. Use for safe operation in hazardous locations.
Energy Improvement Tips
Upgrade your electrical distribution system
Run equipment during "off peak" times as much as possible
Turn off equipment when not in use
Run at higher voltages where possible
For pump applications, regulate flow via motor rpm instead of artificial back pressure or bypass (check out our Frequency Inverters for controlling the speed of 3-phase motors)
 Product Detail
Sort
 Availability
 Price
Direct-Drive Hub-Mount Air Motor, 1/2 hp, 500 to 10,000 rpm
  • Product Type: Hub-mount air motors
  • hp: 1/2
  • Speed range: 500 to 10,000 rpm
EW-07055-72
Usually ships in 5 days.
$246.00 USD/EACH
Qty:
Direct-Drive NEMA type 56 C-Face Air motor, 3/4 hp
  • Product Type: NEMA air motors
  • hp: 3/4
  • Speed range: 350 to 3000 rpm
EW-07055-52
Usually ships in 14 days.
$350.00 USD/EACH
Qty:
Advanced Variable-Speed DC Motor Drive, for 1/4 to 2 hp motors
  • Product Type: DC motor controllers
  • hp: 1/4 to 1 hp @ 115 VAC
    1/2 to 2 hp @ 230 VAC
  • Power: 115/230 VAC
EW-70100-00
Usually ships in 4 days.
$689.00 USD/EACH
Qty:
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1/3 Hp, 1800 RPM
  • Product Type: Three-phase TEFC motors
  • hp: 1/3
  • Speed range: 1800 rpm max
EW-71005-15
Usually ships in 15 days.
$295.00 USD/EACH
Qty:
AC Drive/Frequency Inverter, NEMA 4X, 3 HP, 2.2 kW, 3 In/3 Out; 200-240V
  • Product Type: NEMA 4X frequency inverters
  • hp: 3
  • Power: 200-240 VAC
EW-70022-64
Usually ships in 10 days.
$549.00 USD/EACH
Qty:
AC Drive/Frequency Inverter, NEMA 4X, 0.5 HP, 0.37 kW, 1 or 3 In/3 Out; 200-240V
  • Product Type: NEMA 4X frequency inverters
  • hp: 1/2
  • Power: 200-240 VAC
EW-70022-60
Usually ships in 4 days.
$336.00 USD/EACH
Qty:
AC Drive/Frequency Inverter, NEMA 4X, 3 HP, 2.2 kW, 3 In/3 Out; 400-480V
  • Product Type: NEMA 4X frequency inverters
  • hp: 3
  • Power: 400-480 VAC
EW-70022-69
Usually ships in 10 days.
$581.00 USD/EACH
Qty:
Direct-Drive Hub-Mount Air Motor, 3/4 hp
  • Product Type: Hub-mount air motors
  • hp: 3/4
  • Speed range: 350 to 3000 rpm
EW-07055-74
Usually ships in 8 days.
$185.00 USD/EACH
Qty:
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, 3600 RPM
  • Product Type: Three-phase TEFC motors
  • hp: 1/2
  • Speed range: 3600 rpm max
EW-71005-20
Usually ships in 10 days.
$269.00 USD/EACH
Qty:
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1 Hp, 1800 RPM
  • Product Type: Single-phase stainless steel motors
  • hp: 1
  • Speed range: 1800 rpm max
EW-71012-45
Usually ships in 10 days.
$895.00 USD/EACH
Qty:
AC Drive/Frequency Inverter, NEMA 4X, 1 HP, 0.75 kW, 3 In/3 Out; 480-600V
  • Product Type: NEMA 4X frequency inverters
  • hp: 1
  • Power: 480-600 VAC
EW-70022-70
Usually ships in 10 days.
$466.00 USD/EACH
Qty:
Direct-Drive NEMA Type 56 C-Face Air Motor, 4 hp
  • Product Type: NEMA air motors
  • hp: 4
  • Speed range: 300 to 3000 rpm
EW-07055-76
Usually ships in 9 days.
$503.00 USD/EACH
Qty:
Three-Phase ODP Motor, 56C, 0.33 HP/3600 rpm; 208-230/460 V
  • Product Type: Three-phase ODP motors
  • hp: 1/3
  • Speed range: 3600 rpm max
EW-71011-11
Usually ships in 7 days.
$179.00 USD/EACH
Qty:
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, 1800 RPM
  • Product Type: Three-phase TEFC motors
  • hp: 1/2
  • Speed range: 1800 rpm max
EW-71005-25
Usually ships in 5 days.
$339.00 USD/EACH
Qty:
Stainless Steel NEMA Type C-face Rigid Base Three-phase Motor, 1/3 Hp, 3600 RPM
  • Product Type: Three-phase stainless steel motors
  • hp: 1/3
  • Speed range: 3600 rpm max
EW-71015-10
Usually ships in 10 days.
$495.00 USD/EACH
Qty:
137 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Motors