United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Clean Room
>
Vacuums
> Nilfisk IVT 1000CR Cleanroom Vacuum
Nilfisk IVT 1000CR Cleanroom Vacuum
Collect, contain, and dispose of potent compounds with optional Safe-Pak container
Compatible down to ISO Class 4 (formerly Class 10)
Its compact design, ease of maneuvering, quiet operation, and multistage filtration system make this vacuum ideal for almost any cleanroom environment. The modular constructed, 316L stainless steel (SS) tank and trolley are autoclavable for easy decontamination. Gore-Tex
®
main filter ensures that 99.99% of ultra-fine particulates are retained down to 0.3 microns. ULPA exhaust filter retains 99.99% of particulates at 0.12 microns.
The Safe-Pak is a revolutionary system for safe collection, containment, and disposal of potent compounds. The container is sealed and features a paper bag at the inlet and a HEPA filter on the exhaust. The entire container can be disposed of when full. Ideal for general pharmaceutical cleaning or near tablet presses, capsule-filling machines, and pill de-dusters.
What's Included:
AISI 316 SS trolley and container, and a ULPA exhaust filter. Standard vacuum also includes a Gore- Tex main filter. Safe-Pak model also includes HEPA-filtered disposable Safe-Pak collection container. Order accessories and replacement filters separately.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
SS Cleanroom Vacuum, with Safe-Pak container, 110V
Capacity 3 1/4 Gal,Model 17141208
EW-86564-11
Usually ships in 10 days.
$1,550.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Nilfisk IVT 1000CR Cleanroom Vacuum
SS Cleanroom Vacuum, 110V
Capacity 3 1/4 Gal,Model 17141008
EW-86564-10
Usually ships in 7 days.
$1,550.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Nilfisk
IVT 1000CR Cleanroom Vacuum
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Clean Room
>
Vacuums
> Nilfisk IVT 1000CR Cleanroom Vacuum
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title