Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Availability:
Quantity (mL):
500 (11)
1000 (9)
2500 (9)
100 (1)
20000 (1)
250 (1)
15000 (1)
25 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Nitric Acid

Nitric Acid
Choose from our comprehensive line of Nitric Acid and other Nicotin to Nitrob products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Nitric Acid Certified ACS Plus 2 5 liter

Nitric Acid (Certified ACS Plus) (2.5 liter)

Product Type Chemical Reagents,Purity grade Certified ACS Plus,Form Liquid
EW-88017-74
In stock.
$130.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid 2 5LSafecote

Nitric Acid 2.5LSafecote

Product Type Chemical Reagents,Form Liquid,Quantity mL 2500
EW-88050-70
In stock.
$190.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid Certified ACS Plus 500mL nitric acid reagent acs 500ml

Nitric Acid (Certified ACS Plus) (500mL) nitric acid reagent acs 500ml

Product Type Chemical Reagents,Purity grade Certified ACS Plus,Form Liquid
EW-88017-70
In stock.
$92.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric acid reagent ACS 500ml

Nitric acid, reagent ACS (500ml)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Oxidizing, Corrosive
EW-88362-20
In stock.
$89.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric acid p a 2 5l

Nitric acid p.a. (2.5l)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Corrosive
EW-88123-41
In stock.
$157.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric acid c p 63 1l

Nitric acid c.p., 63% (1l)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Corrosive
EW-88136-84
In stock.
$77.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid Solution 0 1N N 10 Certified 1 liter

Nitric Acid Solution (0.1N (N/10)/Certified) (1 liter)

Product Type Chemical Reagents,Purity grade Certified,Form Liquid
EW-88017-86
In stock.
$107.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid 1 00 Normal 1 L

Nitric Acid, 1.00 Normal (1 L)

Product Type Chemical Reagents,Form Liquid,Quantity mL 1000
EW-86980-90
Usually ships in 10 days.
$34.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid Concentrate Certified 100mL

Nitric Acid Concentrate (Certified) (100mL)

Product Type Chemical Reagents,Purity grade Certified,Form Liquid
EW-88083-95
In stock.
$64.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric acid c p 63 2 5l

Nitric acid c.p., 63% (2.5l)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Corrosive
EW-88136-85
In stock.
$175.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid 1 00 Normal 2 5 L

Nitric Acid, 1.00 Normal (2.5 L)

Product Type Chemical Reagents,Form Liquid,Quantity mL 2500
EW-86984-43
Usually ships in 15 days.
$63.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric acid p a 1l

Nitric acid p.a. (1l)

Product Type Chemical Reagents,Form Liquid,Hazard symbol Corrosive
EW-88123-40
In stock.
$84.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid 6 00 Normal 32 oz

Nitric Acid, 6.00 Normal (32 oz)

Product Type Chemical Reagents,Form Liquid
EW-86980-92
Usually ships in 5 days.
$53.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid Nf 2 1 2L

Nitric Acid Nf 2-1/2L

Product Type Chemical Reagents,Form Liquid,Quantity mL 2500
EW-88053-26
In stock.
$81.00 USD/EACH

Qty:
See all Nitric Acid

Representative photo only Nitric Acid 0 100 Normal N 10 1 L

Nitric Acid, 0.100 Normal (N/10) (1 L)

Product Type Chemical Reagents,Form Liquid,Quantity mL 1000
EW-86980-89
Usually ships in 10 days.
$24.00 USD/EACH

Qty:
See all Nitric Acid

36 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title