Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Availability:
Pore size (micron):
0.45 (2)
0.2 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Life Science > Electrophoresis Products > Filter Membranes > Nitrocellulose membranes

Nitrocellulose membranes

Nitrocellulose membranes

Filter membranes are ideal for use with the gel dryer or semidry blotter. Use pure nitrocellulose membranes for nucleic acid and protein transfers, plaque lifts, and filter hybridization. Binding capacity is 80-100 µg/cm2. Use positively charged nylon membranes for RNA and low-molecular weight DNA transfers without high salt buffers, or whenever a low-protein bonding surface is desirable. Use for multiple DNA or RNA reprobings (not recommended for protein sampling). Nylon membranes are tear-resistant—will not crack or curl.
 Product Detail
Sort
 Availability
 Price
EW-06645-80 Nitrocellulose membranes 0.45um 300cm/roll

Nitrocellulose membranes 0.45um 300cm/roll

Product Type Nitrocellulose Membranes,Filter size 0.45 um,Applications DNA/RNA transfers/reprobing
EW-06645-80
In stock.
$230.00 USD/ROLL

Qty:

Representative photo only Nitrocellulose membranes 0 45um 15x15cm 10 bx

Nitrocellulose membranes 0.45um 15x15cm 10/bx

Product Type Nitrocellulose Membranes,Filter size 0.45 um,Applications DNA/RNA transfers/reprobing
EW-06645-81
Call 1-888-358-4705 for availability.
$113.00 USD/BOX of 10

Qty:

Representative photo only Nitrocellulose membranes 0 2um 300cm roll

Nitrocellulose membranes 0.2um 300cm/roll

Product Type Nitrocellulose Membranes,Filter size 0.45 um,Applications DNA/RNA transfers/reprobing
EW-06645-83
Call 1-888-358-4705 for availability.
$224.00 USD/ROLL

Qty:

3 results (showing 1 - 3 )
Per Page 
Home  >  Life Science > Electrophoresis Products > Filter Membranes > Nitrocellulose membranes

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title