log in or register
Learn more:
[-] Show Less
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Material Testing & Physical Testing > Spectroscopy and Spectrophotometers > Nuclear Magnetic Resonance (NMR) Spectroscopy
Nuclear Magnetic Resonance (NMR) Spectroscopy
Nuclear Magnetic Resonance (NMR) Spectroscopy
A traditional NMR spectrometer (nuclear magnetic resonance) can cost anywhere from hundreds of thousands of dollars, to millions of dollars. By contrast the NMR spectrometer offered through Cole-Parmer consists of one instrument: The world’s first affordable, miniature, easy-to-install and use, yet fully functional FT-NMR spectrometer—the picoSpin 45L. Cole-Parmer is proud to introduce the picoSpin 45L as a revolutionary instrument that greatly expands the availability of a NMR spectrometer. Unlike traditional NMR spectrometers, the picoSpin 45L’s compact size makes it the smallest true FT-NMR spectrometer available. Weighing in at only 10.5 lbs, this solid-state piece of electronics has no moving parts and fits easy into most laboratory settings. However, don’t let its small size fool you. The picoSpin-45L is a complete 45 MHz pulse NMR spectrometer, making it fully capable of monitoring reactions in your lab, pre-screening your samples, monitoring your reaction progress, or supporting many of the other applications for which NMR is known. As a fully functional FT-NMR instrument, it can not only help you identify detailed structural information surrounding chemical compounds, it can also be used for far-ranging applications. If you have a question about the NMR spectrometer offered by Cole-Parmer contact a Technical Expert today.
 Product Detail
Sort
 Availability
 Price
EW-83090-00 2011 Edison Awards Winner - Best New Product: picoSpin 45L Benchtop NMR Spectrometer
picoSpin 45 Benchtop NMR Spectrometer; 115/230 VAC
Product Type NMR Spectrometers,Output FID, JCAMP-DX format,Applications Proton NMR spectroscopy: chemical education and research; analytical testing; quality control; process monitoring and control
EW-83090-00
In stock.
Call for Price
Qty:
EW-83090-50 Capillary Cartridge Replacement for picoSpin-45 Benchtop NMR Spectrometer
Capillary Cartridge Replacement for picoSpin-45 Benchtop NMR Spectrometer
Product Type Capillary Cartridge,Brand picoSpin
EW-83090-50
Call 1-888-358-4705 for availability.
$672.00 USD/EACH
Qty:
Representative photo only Laboratory kit for picoSpin 45 benchtop NMR with 1 16 fittings
EW-83091-52
Usually ships in 8 days.
$250.00 USD/EACH
Qty:
Representative photo only Inlet PEEK filter replacement kit for 1 16 fittings 5PK
EW-83091-53
Call 1-888-358-4705 for availability.
$100.00 USD/PACKAGE
Qty:
Representative photo only Maintenance kit for picoSpin 45 benchtop NMR compatible with 1 32 fittings
EW-83090-53
Usually ships in 11 days or sooner.
$52.50 USD/EACH
Qty:
EW-83090-01 High temperature version of picoSpin
picoSpin-45 HT (High Temperature) Benchtop Spectrometer; 115/230 VAC
Product Type NMR Spectrometers ,Output FID, JCAMP-DX format ,Applications Proton NMR Spectroscopy: chemical education and research; analytical testing; quality control; process monitoring and control
EW-83090-01
Call 1-888-358-4705 for availability.
Call for Price
Qty:
Representative photo only MNOVA Perpetual License for Industry NMR Software for processing analysis simulation and reporting
EW-83090-75
Usually ships in 4 days.
$1,600.00 USD/EACH
Qty:
Representative photo only MNOVA NMR Predict Plug In Perpetual License for Industry NMR Software for processing analysis simulation and reporting
EW-83090-77
Usually ships in 4 days.
$1,350.00 USD/EACH
Qty:
Representative photo only MNOVA Perpetual License for Academia and Government NMR Software for processing analysis simulation and reporting
EW-83090-71
Usually ships in 4 days.
$400.00 USD/EACH
Qty:
EW-83090-02 Fluorine version of picoSpin
picoSpin-45 19F (Fluorine) Benchtop Spectrometer; 115/230 VAC
Product Type NMR Spectrometers ,Output FID, JCAMP-DX format ,Applications Fluorine NMR Spectroscopy; chemical education and research; analytical testing; quality control; process monitoring and control
EW-83090-02
Call 1-888-358-4705 for availability.
Call for Price
Qty:
Representative photo only MNOVA Annual License for Industry NMR Software for processing analysis simulatino and reporting
EW-83090-74
Usually ships in 4 days.
$650.00 USD/EACH
Qty:
Representative photo only MNOVA NMR Predict Plug In Annual License for Industry NMR Software for processing analysis simulation and reporting
EW-83090-76
Usually ships in 4 days.
$550.00 USD/EACH
Qty:
Representative photo only MNOVA Annual License for Academia and Government NMR Software for processing analysis simulation and reporting
EW-83090-70
Usually ships in 4 days.
$150.00 USD/EACH
Qty:
Representative photo only MNOVA NMR Predict Plug In Perpetual License for Academia and Government NMR Software for processing analysis simulation and reporting
EW-83090-73
Usually ships in 4 days.
$350.00 USD/EACH
Qty:
Representative photo only MNOVA NMR Predict Plug In Annual License for Academia and Government NMR Software for processing analysis simulation and reporting
EW-83090-72
Usually ships in 4 days.
$140.00 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
Home  >  Material Testing & Physical Testing > Spectroscopy and Spectrophotometers > Nuclear Magnetic Resonance (NMR) Spectroscopy
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title