Oakton Acorn Thermistor Thermometer
Oakton Acorn Thermistor Thermometer
  • Splashproof sealed membrane keypad makes cleanup easy
  • Minimum and Maximum display
  • Push-button temperature offset calibration adjustment for fine-tuning factory settings
  • HOLD button freezes measured reading
  • Switchable between °C and °F
  • Nonvolatile memory retains data—even when the battery is removed.
  • Auto power-off 17 minutes after last key press
  • Protective rubber boot features a built-in stand.
Ideal for routine temperature measurements, these thermometers offer the convenience and functionality of expensive meters at an economical price. Max/min key momentarily displays maximum or minimum value; or use with HOLD key to update display only on new maximum (minimum) value. Additional features include under range and over range error messages and manual user calibration.
 Product Detail
Sort
 Availability
 Price
Representative photo only Oakton Acorn Temp 5 Thermistor Thermometer With Protective Boot And 35626 50 Probe
Oakton Acorn Temp 5 Thermistor Thermometer With Protective Boot And 35626-50 Probe
Product Type Thermistor Thermometer,Min temperature º F 40,Min temperature º C 40
EW-35626-10
In stock.
$186.95 USD/EACH
Qty:
EW-35626-00 Oakton<small><sup>&reg;</sup></small> Acorn<small><sup>&#153;</sup></small> Temp 4 Thermistor Thermometer With Protective Boot, 400 Series Probe Not Included
Oakton Acorn Temp 4 Thermistor Thermometer With Protective Boot, 400 Series Probe Not Included
Product Type Thermistor Thermometer,Min temperature º F 40,Min temperature º C 40
EW-35626-00
In stock.
$192.95 USD/EACH
Qty:
2 results (showing 1 - 2 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title