log in or register
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Oakton Colorimeter Kits
Oakton Colorimeter Kits
  • Expanded measurement parameters now allow you to measure include chlorine dioxide and ozone!
  • Great for simming pools, spas, drinking water, public utilities, wastewater, and treated water applications
  • Designed to accommodate up to four parameters
  • IP67-rated, waterproof housing floats if accidentally dropped
These Oakton colorimeters feature a large, custom display with clear enunciators. Other features include autoranging, auto-off, and diagnostic error messages for easy use and troubleshooting. Power-saving function allows you to take 3000 readings on four AAA batteries. Colorimeters arrive ready-to-use and factory-calibrated.
 Product Detail
Sort
 Availability
 Price
Representative photo only Oakton C 301 colorimeter pH free and total chlorine
Oakton C-301 colorimeter; pH, free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-30
In stock.
$426.00 USD/EACH
Qty:
EW-35645-20 Oakton<small><sup>&reg;</sup></small> C-201 colorimeter; free and total chlorine.
Oakton C-201 colorimeter; free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-20
In stock.
$386.00 USD/EACH
Qty:
Representative photo only Oakton C 105 colorimeter for ozone
Oakton C-105 colorimeter; for ozone
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-15
Call 1-888-358-4705 for availability.
$337.00 USD/EACH
Qty:
Representative photo only Oakton C 401 colorimeter pH cyanuric acid free and total chlorine
Oakton C-401 colorimeter; pH, cyanuric acid, free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-40
Call 1-888-358-4705 for availability.
$452.00 USD/EACH
Qty:
Representative photo only Oakton C 103 colorimeter for chlorine dioxide
Oakton C-103 colorimeter; for chlorine dioxide
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-13
Call 1-888-358-4705 for availability.
$340.00 USD/EACH
Qty:
5 results (showing 1 - 5 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title