log in or register
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Oakton DO 110 Dissolved Oxygen Meters
Oakton DO 110 Dissolved Oxygen Meters
  • Ideal for testing water quality, wastewater, surface water, groundwater, or drinking water
  • Stores and outputs 100 data sets with temperature in °C or °F
  • Download data via optional RS-232 cable
Meter displays dissolved oxygen (DO) readings in mg/L, ppm, or % saturation with simultaneous temperature display in °F or °C. Independent 100% and 0% calibration points provide high accuracy over the entire range. Increase reading accuracy by entering barometric pressure and salinity—meter automatically calculates offset.
Electrode diagnostics are easily completed with the electrode slope, zero offset, and millivolt display options. Download data via optional RS-232 cable (order separately).
 Product Detail
Sort
 Availability
 Price
Representative photo only Oakton DO 110 Dissolved Oxygen Meter Kit
Oakton DO 110 Dissolved Oxygen Meter Kit
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35640-64
In stock.
$668.00 USD/EACH
Qty:
EW-35640-20 Oakton<small><sup>&reg;</sup></small> DO 110 Dissolved Oxygen Meter with Probe
Oakton DO 110 Dissolved Oxygen Meter with Probe
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35640-20
In stock.
$620.00 USD/EACH
Qty:
EW-35640-22 Oakton<small><sup>&reg;</sup></small> DO 110 Dissolved Oxygen Meter Only
Oakton DO 110 Dissolved Oxygen Meter Only
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35640-22
In stock.
$402.00 USD/EACH
Qty:
Representative photo only Oakton DO 110 Dissolved Oxygen Meter Kit with NIST traceable Calibration
Oakton DO 110 Dissolved Oxygen Meter Kit with NIST-traceable Calibration
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35640-65
Call 1-888-358-4705 for availability.
$767.00 USD/EACH
Qty:
EW-35640-23 Oakton<small><sup>&reg;</sup></small> DO 110 Dissolved Oxygen Meter Only
Oakton DO 110 Dissolved Oxygen Meter Only with NIST-traceable Calibration
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35640-23
Call 1-888-358-4705 for availability.
$501.00 USD/EACH
Qty:
Representative photo only Oakton DO 110 Dissolved Oxygen Meter with Probe and NIST traceable Calibration
Oakton DO 110 Dissolved Oxygen Meter with Probe and NIST-traceable Calibration
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35640-21
Call 1-888-358-4705 for availability.
$719.00 USD/EACH
Qty:
6 results (showing 1 - 6 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title