Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Dissolved Oxygen, BOD, and COD
>
Dissolved Oxygen Meters
>
Dissolved Oxygen Benchtop Meters
> Oakton DO 2700 Benchtop Dissolved Oxygen Meter
Oakton DO 2700 Benchtop Dissolved Oxygen Meter
Designed for optimal performance and versatility
Advanced meter for research grade use with BOD capability
Oakton DO 2700 meter measures dissolved oxygen in mg/L, ppm, or % saturation with simultaneous temperature display in °F or °C. Large LCD features annunciators to aid in meter use and troubleshooting. Large display size makes viewing measurements easier. Meter is encased in a compact housing which takes up a minimum of bench space. Water-resistant membrane keypad and convenient pullout reference guide provide useful features for the lab.
Meter meets GLP requirementsall calibration and measurement data is stored with a time/date stamp. Bundled self-stirring probe and bi-directional RS-232 make this meter ideal for BOD and other advanced dissolved oxygen applications. Nonvolatile memory stores up to 500 data sets to simplify documenting data. Oversized graphic LCD with backlight allows readings under all conditions.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Oakton DO 2700 Benchtop Dissolved Oxygen Meter
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 6-7/8
EW-35416-00
Call 1-888-358-4705 for availability.
$1,845.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton DO 2700 Benchtop Dissolved Oxygen Meter
Oakton DO 2700 Benchtop Dissolved Oxygen Meter with NIST-traceable Calibration
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 6-7/8
EW-35416-01
Call 1-888-358-4705 for availability.
$1,944.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton DO 2700 Benchtop Dissolved Oxygen Meter
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Dissolved Oxygen, BOD, and COD
>
Dissolved Oxygen Meters
>
Dissolved Oxygen Benchtop Meters
> Oakton DO 2700 Benchtop Dissolved Oxygen Meter
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity