Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton ECTestr Pocket Conductivity Testers
> Oakton ECTestr 11+ Multi-Range Pocket Conductivity Testers
Oakton ECTestr 11+ Multi-Range Pocket Conductivity Testers
Operate as either a cup-style or dip-style tester for greater flexibility
Automatically or manually select measuring range
Waterproof, dustproof housings float
Push-button calibration, HOLD function, and error messages
Use these testers for either cup-style or dip-style measurements. For cup style, pour a small amount of your sample directly into the sensor cup until it overflows; for dip style, simply immerse tester tip into sample. Replaceable sensor lets you reuse meter body many times over. Valox
®
housing and stainless steel electrodes for good chemical compatibility.
Note:
Before measuring, calibrate tester as either cup style or dip style (depending on intended method of use).
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Oakton ECTestr 11+ multi-range, cup-style pocket conductivity tester
Product Type Conductivity Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35662-35
In stock.
$112.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton ECTestr 11+ Multi-Range Pocket Conductivity Testers
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton ECTestr Pocket Conductivity Testers
> Oakton ECTestr 11+ Multi-Range Pocket Conductivity Testers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity