United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$300 to $499
(1)
$500 to $799
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Dissolved Oxygen, BOD, and COD
>
Dissolved Oxygen Meters
>
Dissolved Oxygen Handheld Meters
> Oakton Waterproof DO 300 Dissolved Oxygen Meters
Oakton Waterproof DO 300 Dissolved Oxygen Meters
IP67 waterproof and dustproof housings float
Perfect for harsh, rugged field conditions
Store 50 measurements with corresponding temperatures and time
Galvanic probe allows you to take measurements immediately
Dissolved oxygen (DO) readings are displayed in mg/L, ppm, or % saturation simultaneously with temperature values. Adjustable backlit display makes it easy to view readings in low light levels. Calibrate at the independent 100% and 0% calibration points to achieve high accuracy over the entire range. Increase reading accuracy by entering barometric pressure and salinitymeters automatically calculate offset. Setup function displays electrode slope, zero offset, and millivolt. Real-time clock stamps stored readings and calibration data with date and time, making it ideal for Good Laboratory Practice (GLP) standards. Additional features include selectable °C or °F temperature units, Hold function, ready indicator, and auto-off for battery conservation.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Oakton Waterproof DO 300 Meter with Probe
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35641-00
In stock.
$744.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof DO 300 Dissolved Oxygen Meters
Oakton Waterproof DO 300 Meter Only
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35641-01
In stock.
$447.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof DO 300 Dissolved Oxygen Meters
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Dissolved Oxygen, BOD, and COD
>
Dissolved Oxygen Meters
>
Dissolved Oxygen Handheld Meters
> Oakton Waterproof DO 300 Dissolved Oxygen Meters
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title