Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Multiparameter Pocket Testers
> Oakton Waterproof PCSTestr 35 Pocket Multiparameter Testers
Oakton Waterproof PCSTestr 35 Pocket Multiparameter Testers
The ultimate tester for pH, conductivity, TDS, salinity and temperature!
Combine measurement capabilities of three testers in one convenient pocket-sized unit
Up to 5 point pH calibration with NIST or USA autobuffer recognition
Conductivty, TDS, and salinity feature autoranging based on the concentration of your sample
IP67 - rated housing floats if accidentally dropped in water
Great for water, wastewater, laboratory or plant use, these multiparameter Testrs feature long-life pH electrodes with a wide range of sample compatibility. Pin-style conductivity sensors feature stainless steel electrodes for chemical resistance and durability. Replaceable sensor module saves you money by reusing the meter body. Advanced features include auto-shutoff, user-adjustable temperature coefficient, self-adjusting TDS and salinity factor, and low-battery indicator.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Oakton PCSTestr 35 Waterproof Multiparameter pH/conductivity/TDS/salinity pocket tester
Product Type pH Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35425-10
In stock.
$169.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof PCSTestr 35 Pocket Multiparameter Testers
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Multiparameter Pocket Testers
> Oakton Waterproof PCSTestr 35 Pocket Multiparameter Testers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity