Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton Waterproof pH Testr Pocket pH Testers
> Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
Customizable to your applicationuse with any pH electrode with a BNC connector
Versatility of a handheld meter at a pocket price
IP67-rated protects meter from water, dust, and shock while offering chemical resistance
Meter floats—accidental drops into water will not result in lost or damaged meters
Simply choose the electrode best suited for your application, connect the pHTestr BNC 10, calibrate, and measure. Change electrodes to suit your application without having to purchase another meter.
Order electrodes separately.
Large display and tactile keypad allow for easy calibration along with USA and NIST auto-buffer recognition. Hold function freezes reading on the display for easy recording. Additional features include Valox
®
sensor housing, self-diagnostic error messages, a battery-life indicator, and an auto-off function to maximize battery life.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
Product Type pH Tester,Meter style Tester,Dimensions " L 6
EW-35634-14
In stock.
$108.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton Waterproof pH Testr Pocket pH Testers
> Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity