log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Oakton Waterproof pHTestr 10 Pocket pH Tester
Oakton Waterproof pHTestr 10 Pocket pH Tester
  • Versatile, compact design works in a field or laboratory setting
  • A complete meter with key features at an economical price
  • Replaceable, double-junction electrode sensors save time and money
  • IP67-rated housing sits flat on the table and floats in water
Waterproof pHTestr 10 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox® sensor housing resists most chemicals.
The pHTestr 10 has a replaceable sensor for easy maintenance. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage.
The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
Meter includes: protective plastic storage case, lanyard, and batteries.
Meter kit adds: two single-use pouches of pH 4.01, pH 7.00, pH 10.0 buffer, and rinse solution, one sample jar, and sturdy carrying case.
 Product Detail
Sort
 Availability
 Price
Representative photo only Oakton Waterproof pHTestr 10 Pocket pH Tester
Oakton Waterproof pHTestr 10 Pocket pH Tester
Product Type pH Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35634-10
In stock.
$96.00 USD/EACH
Qty:
Representative photo only Oakton Waterproof pHTestr 10 Pocket pH Tester and calibration kit
Oakton Waterproof pHTestr 10 Pocket pH Tester and calibration kit
Product Type pH Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35634-70
In stock.
$123.00 USD/EACH
Qty:
2 results (showing 1 - 2 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title