Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton Waterproof pH Testr Pocket pH Testers
> Oakton Waterproof pHTestr 20 Pocket pH Tester
Oakton Waterproof pHTestr 20 Pocket pH Tester
Versatile, compact design works in a field or laboratory setting
pHTestr 20 features ±0.01 pH accuracysuitable for most ASTM and EPA test procedures.
A complete meter with key features at an economical price
Replaceable, double-junction electrode sensors save time and money
IP67-rated housing sits flat on the table and floats in water
Waterproof pHTestr 20 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox
®
sensor housing resists most chemicals.
The pHTestr 20 has a replaceable sensor for easy maintenance. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar
®
junction and a recessed, glass pH bulb to protect against breakage.
The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
Meter includes:
protective plastic storage case, lanyard, and batteries.
Meter kit adds:
two single-use pouches of pH 4.01, pH 7.00, pH 10.0 buffer, and rinse solution, one sample jar, and sturdy carrying case.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Oakton Waterproof pHTestr 20 (-1.00 to 15.00 pH Range)
Product Type pH Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35634-20
In stock.
$105.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof pHTestr 20 Pocket pH Tester
Oakton Waterproof pHTestr 20 Pocket pH Tester and calibration kit
Product Type pH Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35634-80
In stock.
$147.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof pHTestr 20 Pocket pH Tester
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Pocket Testers
>
Oakton Waterproof pH Testr Pocket pH Testers
> Oakton Waterproof pHTestr 20 Pocket pH Tester
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity