log in or register
Price Range:
Availability:
Temperature compensation:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  pH Meters and Water Quality Products > Multiparameter Meters > Handheld Multiparameter Meters > Oakton Waterproof pH/Con 300 pH/Conductivity Meters
Oakton Waterproof pH/Con 300 pH/Conductivity Meters
Oakton Waterproof pH/Con 300 pH/Conductivity Meters
  • Separate pH and conductivity/TDS probes allow for greater flexibility
  • IP67-rated pH/CON 300 meter floats!
  • Dual display of pH, conductivity or TDS plus temperature (°F/°C selectable)
  • Perform five-point pH and conductivity (TDS) calibration for better accuracy
Features include automatic pH buffer recognition, auto ranging for conductivity/TDS measurements, auto-off, HOLD, and selectable temperature (°C or °F). Meter recalls previous calibration data and pH electrode slope and offset.
 Product Detail
Sort
 Availability
 Price
Representative photo only Oakton Waterproof pH Con 300 Meter with probes
Oakton Waterproof pH/Con 300 Meter with probes
Product Type Multiparameter Meter,Meter style Portable,Dimensions " H 3-1/2
EW-35631-00
In stock.
$821.00 USD/EACH
Qty:
EW-35631-60 Oakton<small><sup>&reg;</sup></small> Waterproof pH/Con 300 Meter Kit
Oakton Waterproof pH/Con 300 Meter Kit
Product Type Multiparameter Meter,Meter style Portable,Dimensions " H 3-1/2
EW-35631-60
In stock.
$922.00 USD/EACH
Qty:
Representative photo only Oakton Waterproof pH Con 300 Meter only
Oakton Waterproof pH/Con 300 Meter only
Product Type Multiparameter Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35631-02
Call 1-888-358-4705 for availability.
$608.00 USD/EACH
Qty:
EW-35631-65 Oakton<small><sup>&reg;</sup></small> Waterproof pH/Con 300 Meter Kit
Oakton Waterproof pH/Con 300 Meter Kit with NIST-traceable Calibration
Product Type Multiparameter Meter,Meter style Portable,Dimensions " H 3-1/2
EW-35631-65
Usually ships in 10 days.
$1,108.00 USD/EACH
Qty:
Representative photo only Oakton Waterproof pH Con 300 Meter with probes and NIST traceable calibration
Oakton Waterproof pH/Con 300 Meter with probes and NIST-traceable calibration
Product Type Multiparameter Meter,Meter style Portable,Dimensions " H 3-1/2
EW-35631-05
Call 1-888-358-4705 for availability.
$1,018.00 USD/EACH
Qty:
5 results (showing 1 - 5 )
Per Page 
Home  >  pH Meters and Water Quality Products > Multiparameter Meters > Handheld Multiparameter Meters > Oakton Waterproof pH/Con 300 pH/Conductivity Meters
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title