log in or register
Brand:
Oakton (5)
Product Type:
Price Range:
Availability:
Waterproof:
Yes (5)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  pH Meters and Water Quality Products > Multiparameter Meters > Handheld Multiparameter Meters > Oakton Waterproof pH/DO 300 pH/Dissolved Oxygen Meter
Oakton Waterproof pH/DO 300 pH/Dissolved Oxygen Meter
Oakton Waterproof pH/DO 300 pH/Dissolved Oxygen Meter
  • Toggle between pH and DO readings with the press of a button
  • Waterproof and dustproof with IP67 rating
  • Reads pH and dissolved oxygen in mg/L, ppm, or % saturation with simultaneous temperature display
Features include five-point pH calibration with auto buffer recognition for +/-0.01 pH accuracy, compensation for salinity and altitude, independent 100% and 0% adjustment calibrations, °C or °F selectable, automatic temperature compensation (ATC), HOLD function, ready indicator, and auto-off. Setup function displays electrode slope, zero offset, and millivolt values. Adjustable backlit display is ideal for taking readings in low-light. The galvanic DO probe allows user to take measurements immediately without warm-up time.
 Product Detail
Sort
 Availability
 Price
EW-35632-60 Oakton<small><sup>&reg;</sup></small> Waterproof pH/DO 300 Meter Kit
Oakton Waterproof pH/DO 300 Meter Kit
Product Type Multiparameter Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35632-60
Usually ships in 6 days or sooner.
$1,489.00 USD/EACH
Qty:
Representative photo only Oakton Waterproof pH DO 300 Meter with probes
Oakton Waterproof pH/DO 300 Meter with probes
Product Type Multiparameter Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35632-00
In stock.
$1,368.00 USD/EACH
Qty:
Representative photo only Oakton Waterproof pH DO 300 Meter Only
Oakton Waterproof pH/DO 300 Meter Only
Meter style Portable,Dimensions " W 3-1/2,Dimensions " H 1-3/4
EW-35632-02
Call 1-888-358-4705 for availability.
$803.00 USD/EACH
Qty:
Representative photo only Oakton Waterproof pH DO 300 Meter with probes and NIST traceable calibrations
Oakton Waterproof pH/DO 300 Meter with probes and NIST-traceable calibrations
Product Type Multiparameter Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35632-03
Call 1-888-358-4705 for availability.
$1,565.00 USD/EACH
Qty:
Representative photo only Oakton Waterproof pH DO 300 Meter Only with NIST traceable calibrations
Oakton Waterproof pH/DO 300 Meter Only with NIST-traceable calibrations
Product Type Multiparameter Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35632-05
Call 1-888-358-4705 for availability.
$1,001.00 USD/EACH
Qty:
EW-35632-62 Oakton<small><sup>&reg;</sup></small> Waterproof pH/DO 300 Meter Kit
Oakton Waterproof pH/DO 300 Meter Kit with NIST-traceable calibrations
Product Type Multiparameter Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35632-62
Usually ships in 6 days.
$1,668.00 USD/EACH
Qty:
6 results (showing 1 - 6 )
Per Page 
Home  >  pH Meters and Water Quality Products > Multiparameter Meters > Handheld Multiparameter Meters > Oakton Waterproof pH/DO 300 pH/Dissolved Oxygen Meter
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title