Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Oakton pH Solution Five Pack

Oakton pH Solution Five Pack

  • Save over 25% compared to ordering separately!
Economical and convenient way to order everything you need to calibrate, clean, and store your pH electrodes. Pack includes one 500-mL bottle each of pH 4.01, 7.00, and 10.00 buffers; one 500-mL bottle of cleaning solution for grease, oil, and protein removal; and one 500-mL bottle of storage solution to properly store your electrode while not in use.
 Product Detail
Sort
 Availability
 Price
Representative photo only Oakton pH Solution Pack one 500 mL bottle each of pH 4 01 7 00 10 00 storage and cleaning solution
EW-05942-15
In stock.
$53.50 USD/PACKAGE

Qty:

1 results (showing 1 - 1 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title