log in or register
Price Range:
Availability:
Capacity (kg):
1 to 10 (10)
Repeatability (g):
Readability (g):
0.1 (8)
0.01 (7)
0.001 (2)
Capacity (g):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Balances & Scales > Toploading Balances > Ohaus Toploading Balances > Ohaus Adventurer Pro Toploading Balances
Ohaus Adventurer Pro Toploading Balances
Ohaus Adventurer Pro Toploading Balances
  • The most complete balance in its class!
  • SmarText™ software guides users through application use and balance setup
  • QuadraStance™ design provides superior footprint stability
  • RS-232 interface allows connection to a PC or printer
Since its introduction, the Ohaus Adventurer Pro has established itself as the industry’s most versatile balance— it counts, it sums, it holds and it delivers traceable results.
Users frustrated by navigating tight spaces will welcome the frameless, flip-top draft shield featured on analytical and high precision models. Designed without a top rail for the glass doors, the draft shield allows easy access into the weighing chamber, helps to eliminate spillage, and reduces waste. To help avoid contamination, all doors, including the front and back glass panels, are easy to remove, and replace. The stainless steel bottom is fast and easy to clean.
Ohaus’ Smart Text balance software utilizes text prompts to guide users through application use and balance setup.
 Product Detail
Sort
 Availability
 Price
EW-11018-12 Ohaus Adventurer™ Toploading Balance, 8100 g x 0.1 g, 115 VAC.  Representative Photo Only
Ohaus Adventurer Pro Toploading Balance, 8100 g x 0.1 g, 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 8100
EW-11018-12
Usually ships in 7 days or sooner.
$1,100.00 USD/EACH
Qty:
EW-11018-00 Ohaus Adventurer™ Toploading Balance, 51 G, 115 VAC.  Representative Photo Only
Ohaus Adventurer Pro Toploading Balance, 51 G, 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 51
EW-11018-00
Usually ships in 13 days.
$1,020.00 USD/EACH
Qty:
EW-11018-02 Ohaus Adventurer™ Toploading Balance, 210 g x 0.01 g, 115 VAC.  Representative Photo Only
Ohaus Adventurer Pro Toploading Balance, 210 g x 0.01 g, 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 210
EW-11018-02
Usually ships in 8 days.
$890.00 USD/EACH
Qty:
EW-11018-08 Ohaus Adventurer™ Toploading Balance, 2100 g x 0.1 g, 115 VAC.  Representative Photo Only
Ohaus Adventurer Pro Toploading Balance, 2100 g x 0.1 g, 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 2100
EW-11018-08
Usually ships in 9 days.
$890.00 USD/EACH
Qty:
EW-11018-06 Ohaus Adventurer™ Toploading Balance, 810 g x 0.01 g, 115 VAC.  Representative Photo Only
Ohaus Adventurer Pro Toploading Balance, 810 g x 0.01 g, 115 VAC
Product Type Toploading Balances,Calibration External,Capacity g 810
EW-11018-06
Usually ships in 9 days.
$1,125.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV213C 210g x 0 001g with InCal Square Draftshield
Ohaus Adventurer Pro Precision Balance AV213C, 210g x 0.001g with InCal & Square Draftshield
Product Type Toploading Balances,Calibration External,Capacity g 210
EW-11600-46
Usually ships in 10 days.
$1,750.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV412C 410g x 0 01g With InCal
Ohaus Adventurer Pro Precision Balance AV412C, 410g x 0.01g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 410
EW-11600-48
Usually ships in 10 days.
$1,090.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV313C 310g x 0 001g With InCal Square Draftshield
Ohaus Adventurer Pro Precision Balance AV313C, 310g x 0.001g With InCal & Square Draftshield
Product Type Toploading Balances,Calibration External,Capacity g 310
EW-11600-47
Usually ships in 10 days.
$1,900.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV812C 810g x 0 01g With InCal
Ohaus Adventurer Pro Precision Balance AV812C, 810g x 0.01g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 810
EW-11600-49
Usually ships in 10 days.
$1,240.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV3102C 3100g x 0 01g With InCal
Ohaus Adventurer Pro Precision Balance AV3102C, 3100g x 0.01g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 3100
EW-11600-52
Usually ships in 10 days.
$1,900.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV212C 210g x 0 01g With InCal
Ohaus Adventurer Pro Precision Balance AV212C, 210g x 0.01g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 210
EW-11600-45
Usually ships in 10 days.
$995.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV2102C 2100g x 0 01g With InCal
Ohaus Adventurer Pro Precision Balance AV2102C, 2100g x 0.01g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 2100
EW-11600-51
Usually ships in 10 days.
$1,700.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV4101C 4100g x 0 1g With InCal
Ohaus Adventurer Pro Precision Balance AV4101C, 4100g x 0.1g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 4100
EW-11600-53
Usually ships in 10 days.
$1,085.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV2101C 2100g x 0 1g With InCal
Ohaus Adventurer Pro Precision Balance AV2101C, 2100g x 0.1g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 2100
EW-11600-50
Usually ships in 10 days.
$1,000.00 USD/EACH
Qty:
Representative photo only Ohaus Adventurer Pro Precision Balance AV8101C 8100g x 0 1g With InCal
Ohaus Adventurer Pro Precision Balance AV8101C, 8100g x 0.1g With InCal
Product Type Toploading Balances,Calibration External,Capacity g 8100
EW-11600-54
Usually ships in 10 days.
$1,205.00 USD/EACH
Qty:
17 results (showing 1 - 15 )
Per Page 
Home  >  Balances & Scales > Toploading Balances > Ohaus Toploading Balances > Ohaus Adventurer Pro Toploading Balances
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title