United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$500 to $575
(1)
$575 to $650
(1)
Readability (lbs):
0.01
(1)
0.02
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Balances & Scales
>
Price Computing and Shipping Scales
> Ohaus RE Standard Price Computing Scale
Ohaus RE Standard Price Computing Scale
The low profile OHAUS RE Series standard price computing scale meets most weighing needs for the cost conscious retailer.
High-grade polymer housing with removable stainless steel pan
Large, easy-to-read backlit LCD displays
Numeric, 32-key positive feedback membrane switch with key-press beeper
Separate tare and zero function keys
Unit price storage up to 100 PLU
24 preset memory keys for quick recall with customizable pull-out card
Total price hold
Pre-pack mode
By-count price computing function
Fractional pricing
Accumulation of total price
Battery operation with auto-shut off and sleep mode power saving features
NTEP approved 3,000d resolution
The RE Series is highly versatile and portable, with its battery operation and price computing features such as: 100 PLU memory, 24 pre-set keys, by-count pricing, and fractional pricing. This makes the RE Series ideal for applications including mobile businesses, retail sale of fruits and vegetables, meats and cheese, delicatessens, confectionery, coffee and tea, and portion control. Easy to operate, easy to clean, and requiring minimum maintenance, the RE Series has everything you need in a standard retail price computing scale.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Ohaus RE Price Computing Compact scale RE-30US , 60lb x 0.02lb, No Tower
Product Type Price Computing Scales,Calibration External,Capacity lbs 30
EW-11600-11
Usually ships in 6 days.
$518.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ohaus RE Standard Price Computing Scale
Ohaus RE Price Computing Compact scale RE-15CUS . 30lb x 0.01lb, With Display Tower
Product Type Price Computing Scales,Calibration External,Capacity lbs 15
EW-11600-10
Usually ships in 6 days.
$584.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ohaus RE Standard Price Computing Scale
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Balances & Scales
>
Price Computing and Shipping Scales
> Ohaus RE Standard Price Computing Scale
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title