Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Meter style:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Orbeco-Hellige Jar Testers

Orbeco-Hellige Jar Testers

  • ASTM D203S compliant method
  • Single motor drives all positions at a consistent speed
  • Choose from laboratory or portable models
  • Available with 4 or 6 positions
Jar testers are designed to test the efficiency of flocculation or preceipitation reagents. The single motor drives all positions at a constant stirring speed. Stirring rod shafts can individually be stopped and height can be adjusted while the system is operating. Choose from laboratory or portable tester models.
Laboratory Jar Testers are available with either four or six positions with a digital display that shows stirring speed and time. Stirring speed is programmable from 10 to 300 rpm, and time from 1 to 999 minutes or 0 to 99 hours. Featuring an illuminated back panel for enhanced visibility. Laboratory units require standard 2-L beakers (order separately,34512-13 4/pk).
Portable Jar Tester is available in a space saving 4-position model with stirring positions around a central lamp. Set stirring speed mechanically to 20, 40, 50, 100 or 120 rpm, and set time for 1 to 30 minutes. Central lamp makes it easier to observe flocculation process. Portable tester requires 1-L beakers (order separately, 34512-11 6/pk).
 Product Detail
Sort
 Availability
 Price
EW-99561-25 PORTABLE JAR TESTER, 4 POSITIONS

Portable Jar Tester, Portable, 4 Positions

Product Type Jar Tester,Meter style Portable,Dimensions " W 12.5
EW-99561-25
Usually ships in 8 days.
$1,820.00 USD/EACH

Qty:

EW-99561-35 LABORATORY JAR TESTER, 4 POSITIONS

Laboratory Jar Tester, 4 Positions

Product Type Jar Tester,Meter style Benchtop,Dimensions " W 10.25
EW-99561-35
Usually ships in 8 days.
$2,450.00 USD/EACH

Qty:

EW-99561-45 LABORATORY JAR TESTER, 6 POSITIONS

Laboratory Jar Tester, 6 Positions

Product Type Jar Tester,Meter style Benchtop,Dimensions " W 10.25
EW-99561-45
Usually ships in 8 days.
$2,580.00 USD/EACH

Qty:

3 results (showing 1 - 3 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title