log in or register
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Masterflex Peristaltic Pumps > Masterflex Tubing and Pump Accessories > Overlook Industries OneShot Single-Use Filler Nozzles
Overlook Industries OneShot Single-Use Filler Nozzles
Overlook Industries OneShot Single-Use Filler Nozzles
  • Reduce filler line downtime and risk of cross-contamination
  • Lower costs for cleaning, sterilization, and validation
  • Provide enhanced flexibility in batch scheduling and product development
  • Dimensional tolerances of +/-0.05% for assured repeatability
These nozzles combine the ease of single-use plastic components with the accuracy and reliability of stainless steel filler needles. Tight dimensional tolerances help to ensure precision in your dispensing applications. Nozzles feature a 316L stainless steel needle and a base with hose barb adapter. Materials meet FDA, USDA and USP Class VI requirements. All filler nozzles are sterilizable by ethylene oxide, autoclaving, or gamma irradiation (note: polycarbonate will turn a greenish-yellow hue when gamma irradiated).
 Product Detail
Sort
 Availability
 Price
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 1 8
Disposable filler nozzle, SS needle and polycarbonate base, 1/8
Product Type Biopharma fittings,Tubing ID in. 0.125,Material Polycarbonate
EW-30619-07
In stock.
$105.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 1 16
Disposable filler nozzle, SS needle and polycarbonate base, 1/16
Product Type Biopharma fittings,Tubing ID in. 0.0625,Material Polycarbonate
EW-30619-01
In stock.
$105.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 3 16
Disposable filler nozzle, SS needle and polycarbonate base, 3/16
Product Type Biopharma fittings,Tubing ID in. 0.1875,Material Polycarbonate
EW-30619-02
In stock.
$95.00 USD/EACH
Qty:
EW-30619-08 Disposable filler nozzle, SS needle and polycarbonate base, 1/4
Disposable filler nozzle, SS needle and polycarbonate base, 1/4
Product Type Biopharma fittings,Tubing ID in. 0.25,Material Polycarbonate
EW-30619-08
In stock.
$97.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needleand polycarbonate base 3 8
Disposable filler nozzle, SS needleand polycarbonate base, 3/8
Product Type Biopharma fittings,Tubing ID in. 0.375,Material Polycarbonate
EW-30619-09
In stock.
$105.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 1 32
Disposable filler nozzle, SS needle and polycarbonate base, 1/32
Product Type Biopharma fittings,Tubing ID in. 0.03125,Material Polycarbonate
EW-30619-06
Call 1-888-358-4705 for availability.
$115.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 5 16
Disposable filler nozzle, SS needle and polycarbonate base, 5/16
Product Type Biopharma fittings,Tubing ID in. 0.3125,Material Polycarbonate
EW-30619-03
Usually ships in 14 days.
$97.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 5 8
Disposable filler nozzle, SS needle and polycarbonate base, 5/8
Product Type Biopharma fittings,Tubing ID in. 0.625,Material Polycarbonate
EW-30619-10
Usually ships in 14 days.
$105.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 3 4
Disposable filler nozzle, SS needle and polycarbonate base, 3/4
Product Type Biopharma fittings,Tubing ID in. 0.75,Material Polycarbonate
EW-30619-05
Usually ships in 14 days.
$99.00 USD/EACH
Qty:
Representative photo only Disposable filler nozzle SS needle and polycarbonate base 1 2
Disposable filler nozzle, SS needle and polycarbonate base, 1/2
Product Type Biopharma fittings,Tubing ID in. 0.5,Material Polycarbonate
EW-30619-04
In stock.
$95.00 USD/EACH
Qty:
10 results (showing 1 - 10 )
Per Page 
Home  >  Masterflex Peristaltic Pumps > Masterflex Tubing and Pump Accessories > Overlook Industries OneShot Single-Use Filler Nozzles
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title