log in or register
Availability:
Material:
HDPE (19)
Max temperature:
Volume (gal.):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fluid & Liquid Handling > Tanks and Drums > Plastic Tanks and Drums > PE Cylindrical Tanks with Covers
PE Cylindrical Tanks with Covers
PE Cylindrical Tanks with Covers
  • Seamless tanks are rotation-molded
Cylindrical tanks have rounded corners and an outside rim flange for strength and ease of handling. To order larger tanks, or tanks for special applications, call our Application Specialists for more information.
Tanks with 30-gallon capacity or less have a raised-center bottom for complete drainage. Tanks with 55-gallon capacity or more have a spoked bottom.
High-density polyethylene (HDPE) tanks come with or without a needle-type polypropylene spigot. Spigot has 1-1/8"-12 female screw thread. Spigot is mounted on a welded polyethylene boss and accepts 5/8" ID tubing.
Polypropylene (PP) tanks offer good stress-crack and chemical resistance. Maximum service temperature up to 104°C (220°F).
 Product Detail
Sort
 Availability
 Price
Representative photo only Cylidrical Tank with Cover PP 30 Gal
Cylidrical Tank with Cover, PP, 30 Gal.
Product Type Cylindrical Tanks,Volume gal. 30,Material Polypropylene
EW-06317-73
In stock.
$266.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover HDPE 15 Gal
Cylidrical Tank with Cover, HDPE, 15 Gal.
Product Type Cylindrical Tanks,Volume gal. 15,Material HDPE
EW-06317-65
In stock.
$122.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover PP 55 Gal
Cylidrical Tank with Cover, PP, 55 Gal.
Product Type Cylindrical Tanks,Volume gal. 55,Material Polypropylene
EW-06317-78
Usually ships in 5 days.
$355.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover PP 100 Gal
Cylidrical Tank with Cover, PP, 100 Gal.
Product Type Cylindrical Tanks,Volume gal. 100,Material Polypropylene
EW-06317-13
Usually ships in 15 days.
$754.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover HDPE 200 Gal
Cylidrical Tank with Cover, HDPE, 200 Gal.
Product Type Cylindrical Tanks,Volume gal. 200,Material HDPE
EW-06317-31
Call 1-888-358-4705 for availability.
$728.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover HDPE 100 Gal
Cylidrical Tank with Cover, HDPE, 100 Gal.
Product Type Cylindrical Tanks,Volume gal. 100,Material HDPE
EW-06317-10
Usually ships in 8 days.
$499.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover HDPE 55 Gal
Cylidrical Tank with Cover, HDPE, 55 Gal.
Product Type Cylindrical Tanks,Volume gal. 55,Material HDPE
EW-06317-75
Usually ships in 5 days.
$266.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover and Spigot HDPE 55 Gal 1 4 Wall
Cylidrical Tank with Cover and Spigot, HDPE, 55 Gal., 1/4" Wall
Product Type Cylindrical Tanks,Volume gal. 55,Material HDPE
EW-06317-76
Usually ships in 6 days.
$303.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover HDPE 30 Gal
Cylidrical Tank with Cover, HDPE, 30 Gal.
Product Type Cylindrical Tanks,Volume gal. 30,Material HDPE
EW-06317-70
In stock.
$194.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover and Spigot HDPE 30 Gal 3 16 Wall
Cylidrical Tank with Cover and Spigot, HDPE, 30 Gal., 3/16" Wall
Product Type Cylindrical Tanks,Volume gal. 30,Material HDPE
EW-06317-71
In stock.
$232.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover and Spigot HDPE 15 Gal 3 16 Wall
Cylidrical Tank with Cover and Spigot, HDPE, 15 Gal., 3/16" Wall
Product Type Cylindrical Tanks,Volume gal. 15,Material HDPE
EW-06317-66
In stock.
$163.00 USD/EACH
Qty:
EW-06317-56 Cylidrical Tank with Cover and Spigot, HDPE, 7-1/2 Gal., 3/16" Wall
Cylidrical Tank with Cover and Spigot, HDPE, 7-1/2 Gal., 3/16" Wall
Product Type Cylindrical Tanks,Volume gal. 7.5,Material HDPE
EW-06317-56
In stock.
$131.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover and Spigot HDPE 10 Gal 3 16 Wall
Cylidrical Tank with Cover and Spigot, HDPE, 10 Gal., 3/16" Wall
Product Type Cylindrical Tanks,Volume gal. 10,Material HDPE
EW-06317-61
In stock.
$134.00 USD/EACH
Qty:
Representative photo only Cylidrical Tank with Cover HDPE 275 Gal
Cylidrical Tank with Cover, HDPE, 275 Gal.
Product Type Cylindrical Tanks,Volume gal. 275,Material HDPE
EW-06317-41
Usually ships in 15 days.
$967.00 USD/EACH
Qty:
EW-06317-60 Cylidrical Tank with Cover, HDPE, 10 Gal.
Cylidrical Tank with Cover, HDPE, 10 Gal.
Product Type Cylindrical Tanks,Volume gal. 10,Material HDPE
EW-06317-60
In stock.
$99.00 USD/EACH
Qty:
30 results (showing 1 - 15 )
Per Page 
Home  >  Fluid & Liquid Handling > Tanks and Drums > Plastic Tanks and Drums > PE Cylindrical Tanks with Covers
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title