log in or register
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Gaskets > Sanitary Gaskets > Sanitary Screen Gaskets > PTFE screen gaskets
PTFE screen gaskets
PTFE screen gaskets
  • Protect your pump, valves and other components in fluid line
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, FDA CFR Title 21 Section 177.2600, USDA, 3-A and cGMP criteria. All gaskets are animal-derived ingredient free. Each gasket sold individually.
PTFE gaskets offer the best chemical compatibility and have exceptional resistance to high temperatures, corrosion, and stresscracking. PTFE gaskets will meet the highest demands in pharmaceutical, food and chemical applications. Use gaskets at temperatures from –450 to 550deg F (–268 to 288degC).
 Product Detail
Sort
 Availability
 Price
Representative photo only PTFE Gasket 40 MESH Size 1 1 2 Tri Clamp
PTFE Gasket, 40 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material PTFE
EW-30538-46
Usually ships in 5 days.
$86.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 100 MESH Size 1 Tri Clamp
PTFE Gasket, 100 MESH, Size 1" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1,Material PTFE
EW-30538-84
Usually ships in 6 days.
$86.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 100 MESH Size 1 1 2 Tri Clamp
PTFE Gasket, 100 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material PTFE
EW-30538-86
Usually ships in 6 days.
$86.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 60 MESH Size 1 1 2 Tri Clamp
PTFE Gasket, 60 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material PTFE
EW-30538-66
Call 1-888-358-4705 for availability.
$86.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 40 MESH Size 1 Tri Clamp
PTFE Gasket, 40 MESH, Size 1" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1,Material PTFE
EW-30538-44
Usually ships in 4 days.
$86.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 20 MESH Size 1 1 2 Tri Clamp
PTFE Gasket, 20 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material PTFE
EW-30538-26
Usually ships in 6 days.
$86.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 10 MESH Size 2 Tri Clamp
PTFE Gasket, 10 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material PTFE
EW-30538-08
Usually ships in 7 days.
$95.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 100 MESH Size 2 Tri Clamp
PTFE Gasket, 100 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material PTFE
EW-30538-88
Usually ships in 6 days.
$95.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 100 MESH Size 1 2 Tri Clamp
PTFE Gasket, 100 MESH, Size 1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 0.5,Material PTFE
EW-30538-80
Call 1-888-358-4705 for availability.
$94.00 USD/EACH
Qty:
EW-30538-68 PTFE Gasket, 60 MESH, Size 2" Tri-Clamp
PTFE Gasket, 60 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material PTFE
EW-30538-68
Usually ships in 5 days.
$95.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 20 MESH Size 2 Tri Clamp
PTFE Gasket, 20 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material PTFE
EW-30538-28
Usually ships in 6 days.
$95.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 40 MESH Size 2 Tri Clamp
PTFE Gasket, 40 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material PTFE
EW-30538-48
In stock.
$95.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 10 MESH Size 1 Tri Clamp
PTFE Gasket, 10 MESH, Size 1" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1,Material PTFE
EW-30538-04
Usually ships in 7 days.
$86.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 60 MESH Size 1 2 Tri Clamp
PTFE Gasket, 60 MESH, Size 1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 0.5,Material PTFE
EW-30538-60
Usually ships in 3 days.
$94.00 USD/EACH
Qty:
Representative photo only PTFE Gasket 10 MESH Size 1 1 2 Tri Clamp
PTFE Gasket, 10 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material PTFE
EW-30538-06
Call 1-888-358-4705 for availability.
$86.00 USD/EACH
Qty:
19 results (showing 1 - 15 )
Per Page 
Home  >  Fittings > Gaskets > Sanitary Gaskets > Sanitary Screen Gaskets > PTFE screen gaskets
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title