Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 

Pa to Pentae

Pa to Pentae
Choose from our comprehensive line of Pa to Pentae and other Chemicals P products.
 Product Detail
Sort
 Availability
 Price

Paraformaldehyde 90-92% (2.5kg) CAS 30525-89-4

  • Product Type: Organic Chemicals
  • CAS Number: 30525-89-4
  • Empirical formula: (CH2O)n
EW-88180-71
Usually ships in 5 days.
$95.25 USD/EACH
$86.99 USD/EACH

Qty:

Paraformaldehyde 96% (500g) CAS 30525-89-4

  • Product Type: Organic Chemicals
  • CAS Number: 30525-89-4
  • Form: Solid
EW-88353-82
Usually ships in 2 days.
$45.00 USD/EACH

Qty:

Acros Organics Pectin (Pract), CAS 9000-69-5; 250 g

  • Product Type: Organic Chemicals
  • CAS Number: 9000-69-5
  • Form: Solid
EW-88353-92
Usually ships in 2 days.
$106.00 USD/EACH

Qty:

PAN (Certified) (5g) CAS 85-85-8 (1-(2-Pyridylazo)-2-naphthol)

  • Product Type: Organic Chemicals
  • Purity grade: Certified
  • CAS Number: 85858
EW-88079-33
Usually ships in 2 days.
$350.00 USD/EACH

Qty:

Papanicolaou Stain, Gill's Modified OG-6 Formula, Counterstain (1 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 64-17-5, 67-56-1, 1936-15-8, 12067-99-1 and 7732-18-5
  • Form: Liquid
EW-86981-04
Available for order
or call 1-888-358-4705 to order
$66.75 USD/EACH

Qty:

Palladium on 1/8" alumina pellets 0.5% Pd (250g)

  • Product Type: Inorganic Chemicals
  • CAS Number: 7440-05-3
  • Form: Pellets
EW-88207-64
Usually ships in 2 days.
$577.00 USD/EACH

Qty:

Palladium atomic absorption standard solution 1 mg/ml Pd in 10% HCl (100 mL)

  • Product Type: Inorganic Chemicals
  • CAS Number: 7440-05-3
  • Form: Liquid
EW-88209-19
Usually ships in 2 days.
$86.75 USD/EACH
$66.99 USD/EACH

Qty:

Papanicolaou Stain, EA-65 Formula, Counterstain (1 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 10114-58-6, 17372-87-1, 64-17-5, 5141-20-8, 554-13-2, 67-56-1, 12067-99-1 and 7732-18-5
  • Form: Liquid
EW-86981-01
Available for order
or call 1-888-358-4705 to order
$72.25 USD/EACH

Qty:

Papanicolaou Stain, Gill's Modified OG-6 Formula, (500 mL)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 64-17-5, 67-56-1, 1936-15-8, 12067-99-1 and 7732-18-5
  • Form: Liquid
EW-86974-22
Usually ships in 12 days.
$43.50 USD/EACH

Qty:

Palladium on 4- to 8-mesh carbon 1% Pd (25g)

  • Product Type: Inorganic Chemicals
  • Form: Solid
  • Formula weight: 106.42
EW-88207-65
Usually ships in 14 days.
$135.00 USD/EACH

Qty:

Papanicolaou Stain, EA-65 Formula, Counterstain (4 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 10114-58-6, 17372-87-1, 64-17-5, 5141-20-8, 554-13-2, 67-56-1, 12067-99-1 and 7732-18-5
  • Form: Liquid
EW-86986-57
Usually ships in 7 days.
$155.00 USD/EACH

Qty:

Papanicolaou Stain, OG-6 Formula, Modified, Counterstain (1 L)

  • Product Type: Inorganic Chemicals
  • CAS Number: Includes 64-17-5, 67-56-1, 1936-15-8, 12067-99-1 and 7732-18-5
  • Form: Liquid
EW-86981-02
Usually ships in 14 days.
$56.25 USD/EACH

Qty:

Para Red, dye content ca. 95% (25g) CAS 6410-10-2

  • Product Type: Organic Chemicals
  • CAS Number: 6410-10-2
  • Form: Solid
EW-88144-58
Usually ships in 2 days.
$61.25 USD/EACH
$54.99 USD/EACH

Qty:

Paraformaldehyde 90-92% (1kg) CAS 30525-89-4

  • Product Type: Organic Chemicals
  • CAS Number: 30525-89-4
  • Form: Solid
EW-88180-70
Usually ships in 2 days.
$87.00 USD/EACH

Qty:

n-Pentadecane 99% (250 mL) CAS 629-62-9 (Liquid)

  • Product Type: Organic Chemicals
  • CAS Number: 629-62-9
  • Form: Liquid
EW-88131-54
Usually ships in 2 days.
$149.00 USD/EACH

Qty:

34 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#