Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Palladium on carbon (10%) (50% wet with water for safety)

Palladium on carbon (10%) (50% wet with water for safety)

Palladium on carbon (10%) (50% wet with water for safety)
Choose from our comprehensive line of Palladium on carbon (10%) (50% wet with water for safety) and other Pa to Pentae products.
 Product Detail
Sort
 Availability
 Price
Palladium on carbon 10 50 wet with water for safety 10g  (Representative photo only)
EW-88360-27
In stock.
$215.00 USD/EACH

Qty:

Palladium on carbon 10 50 wet with water for safety 25g  (Representative photo only)
EW-88360-28
In stock.
$520.00 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 
Home  >  Palladium on carbon (10%) (50% wet with water for safety)