log in or register
2,600 pages
of answers
FREE
2013/14 Cole-Parmer
Sourcebook
More than 4 pages of disposable respirators, plus more respiratory protection products are contained in the 2009/2010 general catalog.
Request your copy today.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Particulate Respirators
Particulate Respirators
Cole-Parmer carries a wide selection of Disposable Particulate Respirators that are NIOSH certified, helping you comply with OSHA CFR 1910.134. According to CFR 1910.134(d), employers are required to select and provide an appropriate respirator based on an employer evaluation of the respiratory hazard(s) to which the worker is exposed, and on workplace and user factors that affect respirator performance and reliability. All respirators should be NIOSH-certified and must be used in compliance with the conditions of certification.
Determine the following when choosing a particulate respirator:
  • Are there oil particles (lubricants, cutting fluids, glycerine, etc.) present?
  • Level of particulate removal desired
  • Series Efficiency
    95% 99% 99.97%
    N (not for use with oil particulates) N95 N99 N100
    R (oil resistant, but may require time limits) R95 R99 R100
    P (oil proof) P95 P99 P100
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
EW-86401-00 3M<small><sup>&reg;</sup></small> Three-Panel Particulate respirator, N95, Box of 20
3M Three-Panel Particulate respirator, N95, Box of 20
Product Type Particulate Respirators,Color White,Qty/bx 20
EW-86401-00
In stock.
$37.00 USD/BOX
Qty:
EW-40111-02 3M<small><sup>&reg;</sup></small> Particulate Respirators, N95, Standard Particulate, Box of 20
3M Particulate Respirators, N95, Standard Particulate, Box of 20
Product Type Particulate Respirators,Color White,Qty/bx 20
EW-40111-02
In stock.
$20.00 USD/BOX of 20
Qty:
EW-40111-26 Specialty 3M<small><sup>&reg;</sup></small> Particulate Disposable Respirators, P95, Standard, organic vapor, Cool Flow valve
EW-40111-26
In stock.
$55.00 USD/BOX of 10
Qty:
EW-81660-80 Sperian<small><sup>&reg;</sup></small> Saf-T-Fit<small><sup>&reg;</sup></small> Nuisance Dust Mask, Box of 50
Sperian Saf-T-Fit Nuisance Dust Mask, Box of 50
Product Type Particulate Respirators,Color White,Qty/bx 50
EW-81660-80
In stock.
$8.25 USD/BOX
Qty:
Representative photo only Specialty 3M Particulate Disposable Respirators N100 Cool Flow valve
Specialty 3M Particulate Disposable Respirators, N100, Cool Flow valve
Product Type Particulate Respirators,Color White,Qty/bx 20
EW-40111-14
Usually ships in 14 days.
$196.00 USD/BOX of 20
Qty:
EW-86401-10 3M<small><sup>&#153;</sup></small>Three-Panel Particulate respirator with exhalation valve, N95, Box of 10
3MThree-Panel Particulate respirator with exhalation valve, N95, Box of 10
Product Type Particulate Respirators,Color White,Qty/bx 10
EW-86401-10
In stock.
$37.00 USD/BOX
Qty:
EW-40111-15 Specialty 3M<small><sup>&reg;</sup></small> Particulate Disposable Respirators, P100, Cool Flow valve
Specialty 3M Particulate Disposable Respirators, P100, Cool Flow valve
Product Type Particulate Respirators,Color White,Brand 3M
EW-40111-15
In stock.
$13.25 USD/EACH
Qty:
Representative photo only Sperian Particulate Respirators N95 Organic Vapor Exhalation Valve Small Box of 10
EW-81660-18
Usually ships in 9 days.
$42.00 USD/BOX
Qty:
Representative photo only 3M Particulate Respirators N95 Cool flow valve Box of 10
3M Particulate Respirators, N95, Cool-flow valve, Box of 10
Product Type Particulate Respirators,Color White,Qty/bx 10
EW-40111-04
In stock.
$27.00 USD/BOX of 10
Qty:
EW-40111-20 Specialty 3M<small><sup>&reg;</sup></small> Particulate Disposable Respirators, R95, Standard, acid gas
Specialty 3M Particulate Disposable Respirators, R95, Standard, acid gas
Product Type Particulate Respirators,Color Gray,Qty/bx 20
EW-40111-20
In stock.
$83.00 USD/BOX of 20
Qty:
EW-40111-22 Specialty 3M Particulate Disposable Respirators, R95, Standard, organic vapor
Specialty 3M Particulate Disposable Respirators, R95, Standard, organic vapor
Product Type Particulate Respirators,Color Gray,Qty/bx 20
EW-40111-22
In stock.
$86.00 USD/BOX of 20
Qty:
Representative photo only Specialty 3M Particulate Disposable Respirators P95 Standard acid gas Cool Flow valve
Specialty 3M Particulate Disposable Respirators, P95, Standard, acid gas, Cool Flow valve
Product Type Particulate Respirators,Color White,Qty/bx 10
EW-40111-24
In stock.
$56.00 USD/BOX of 10
Qty:
EW-86214-10 Moldex<small><sup>&reg;</sup></small> N95 Respirators with Exhalation Valve, Small, N95, Box of 10
Moldex N95 Respirators with Exhalation Valve, Small, N95, Box of 10
Product Type Particulate Respirators,Color Blue,Qty/bx 10
EW-86214-10
In stock.
$28.00 USD/BOX of 10
Qty:
Representative photo only Sperian Saf T Fit Plus Particulate Respirators P95 Extra Large Box of 20
EW-81660-08
Usually ships in 9 days.
$46.00 USD/BOX
Qty:
Representative photo only Sperian Saf T Fit Particulate Respirators N95 Standard Small Box of 20
Sperian Saf-T-Fit Particulate Respirators; N95, Standard, Small, Box of 20
Product Type Particulate Respirators,Color White,Qty/bx 20
EW-81660-00
In stock.
$23.50 USD/BOX of 20
Qty:
35 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title