Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Ohaus (451)
Product Type:
Availability:
In Stock (303)
Capacity (mL):
0 - 20 (20)
Calibration:
External (872)
Internal (230)
Capacity (g):
Capacity (kg):
1 to 10 (207)
20 to 50 (108)
Capacity (lbs):
1 to 10 (48)
Material:
PTFE (6)
Membrane material:
PTFE (4)
Pore size (micron):
0.2 (11)
CE Compliance:
Yes (5)
Effective filtration area:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Shop by Industry > Pharmaceutical Industry Products

Pharmaceutical Industry Products

Pharmaceutical Industry Products

Because of its regulations and standards, the pharmaceutical industry has many specific requirements for documentation and purity. Because of these strict standards it is important that companies in the pharmaceutical industry buy their supplies from industry experts who know their needs and can help them. Cole-Pamer understands those needs and works to provide products that include the necessary paperwork and compliance with industry and governmental standards.
Whether for pharma labs and research, ramp-up operations, or full-scale production for a pharmaceutical industry plant or bioprocessing plant, Cole-Parmer can supply exactly the right product. A wide range of laboratory and process products, including several single-use products, can meet your needs. Look for applicable standards met: United States Pharmacopeia (USP), Food and Drug Administration (FDA), GLP/GMP. Look for fluid handling products; labware, lab instrumentation, and lab equipment; chemicals and reagents; and laboratory consumables. All consumables in particular indicate with which standards they comply, so you can rest assured that your process is protected from contamination or other extraneous factors. If you have questions about the products available for companies in the pharmaceutical industry contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
EW-10000-15 Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 115 VAC.  Representative Photo Only

Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 115 VAC

Product Type Analytical Balances,Calibration Internal,Capacity g 120
EW-10000-15
In stock.
$1,540.00 USD/EACH

Qty:

EW-01017-50 Representative Photo Only.  Aluminum Crimpled-Walled Dishes with Tab, 57mm Dia x 5/8

Aluminum Crimpled-Walled Dishes with Tab, 57mm Dia x 5/8" D, 1.3 gr., 60 ml Capacity, 1000/cs

Product Type Weighing Dish,Capacity mL 60,Material aluminum
EW-01017-50
In stock.
$96.00 USD/CASE of 1000

Qty:
See all

EW-98553-10 Representative Photo Only.  PP Miniature Check Valve, 1/8" hose barb, Viton<small><sup>&reg;</sup></small> diaphragm

PP Miniature Check Valve, 1/8" hose barb, Viton diaphragm

Product Type Check Valves (manual),Cv 0.11,Material polypropylene
EW-98553-10
In stock.
$9.75 USD/EACH

Qty:

EW-10000-16 Cole-Parmer<small><sup>&reg;</sup></small> Symmetry<small><sup>&reg;</sup></small> PA-Analytical Balance, 220g x 0.0001g -- 115 VAC.  Representative Photo Only

Cole-Parmer Symmetry PA-Analytical Balance, 220g x 0.0001g -- 115 VAC

Product Type Analytical Balances,Calibration Internal,Capacity g 220
EW-10000-16
Call 1-888-358-4705 for availability.
$1,820.00 USD/EACH

Qty:

EW-10000-14 Cole-Parmer<small><sup>&reg;</sup></small> Symmetry<small><sup>&reg;</sup></small> PR-Precision Toploading Balance, 4200 g x 0.01g, -- 115 VAC.  Representative Photo Only

Cole-Parmer Symmetry PR-Precision Toploading Balance, 4200 g x 0.01g, -- 115 VAC

Product Type Toploading Balances,Calibration External,Capacity g 4200
EW-10000-14
In stock.
$968.00 USD/EACH

Qty:

EW-01017-51 Representative Photo Only.  Aluminum Crimpled-Walled Dishes with Tab, 43mm Dia x 1/2

Aluminum Crimpled-Walled Dishes with Tab, 43mm Dia x 1/2" D, 1 gr., 20 ml Capacity, 1000/cs

Product Type Weighing Dish,Capacity mL 20,Material aluminum
EW-01017-51
In stock.
$93.00 USD/CASE of 1000

Qty:
See all

EW-11500-00 Marble Balance Table

Marble Balance Table

Product Type Accessory,Model TABLE
EW-11500-00
Call 1-888-358-4705 for availability.
$1,415.00 USD/EACH

Qty:

Representative photo only 60 mL B D plastic disposable syringe luer lock tip 40 box

60 mL B-D plastic disposable syringe, luer-lock tip, 40 / box

Material Polycarbonate,Product Type Chromatography syringe,Capacity mL 60
EW-07940-30
Usually ships in 15 days.
$54.00 USD/BOX

Qty:

Representative photo only Square Weigh Boat medium white 3 1 2 top i d x 1 d 100 mL weigh capacity 500 pk

Square Weigh Boat, medium, white, 3-1/2" top i.d. x 1"d 100 mL weigh capacity 500/pk

Product Type Weighing Dish,Capacity mL 100,Color white,Material antistatic polystyrene
EW-01017-15
In stock.
$55.00 USD/PACKAGE of 500

Qty:

EW-11018-12 Ohaus Adventurer™ Toploading Balance, 8100 g x 0.1 g, 115 VAC.  Representative Photo Only

Ohaus Adventurer Pro Toploading Balance, 8100 g x 0.1 g, 115 VAC

Product Type Toploading Balances,Calibration External,Capacity g 8100
EW-11018-12
Usually ships in 7 days or sooner.
$1,100.00 USD/EACH

Qty:

EW-10000-26 Cole-Parmer<small><sup>&reg;</sup></small> Symmetry<small><sup>&reg;</sup></small> PA-Analytical Balance, 220g x 0.0001g -- 220 VAC.  Representative Photo Only

Cole-Parmer Symmetry PA-Analytical Balance, 220g x 0.0001g -- 220 VAC

Product Type Analytical Balances,Calibration Internal,Capacity g 220
EW-10000-26
In stock.
$1,820.00 USD/EACH

Qty:

Representative photo only Hexagonal Weigh Dish Extra Large 5 1 2 o d X 1 1 4 d 350ml capacity 500 pk

Hexagonal Weigh Dish, Extra-Large, 5-1/2"o.d. X 1-1/4"d, 350ml capacity, 500/pk

Product Type Weighing Dish,Capacity mL 350,Material antistatic polystyrene
EW-01018-16
In stock.
$117.00 USD/PACKAGE of 500

Qty:

EW-10000-12 Cole-Parmer<small><sup>&reg;</sup></small> Symmetry<small><sup>&reg;</sup></small> PR-Precision Toploading Balance, 410 g x 0.001g -- 115 VAc.  Representative Photo Only

Cole-Parmer Symmetry PR-Precision Toploading Balance, 410 g x 0.001g -- 115 VAc

Product Type Toploading Balances,Calibration External,Capacity g 410
EW-10000-12
In stock.
$949.00 USD/EACH

Qty:

EW-01002-81 Ohaus MB Moisture Determination Balance, Mb23 110 G X 0.01 G 120VAC

Ohaus MB Moisture Determination Balance, Mb23 110 G X 0.01 G 120VAC

Product Type Moisture Balances,Calibration External,Capacity g 110
EW-01002-81
In stock.
$2,595.00 USD/EACH

Qty:

EW-11010-50 Ohaus Scout® Pro Portable Balance, 200 g, 115 VAC

Ohaus Scout Pro Portable Balance, 200 g, 115 VAC

Product Type Portable Balances,Calibration External,Capacity g 200
EW-11010-50
In stock.
$387.00 USD/EACH

Qty:

2016 results (showing 1 - 15 )
Per Page 
Home  >  Shop by Industry > Pharmaceutical Industry Products

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title