United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Surface Testing
>
Thickness Gauges
> Phase II Digital Ultrasonic Thickness Gauge
Phase II Digital Ultrasonic Thickness Gauge
Packed with features typically found only on high-end models
Stores up to 20 groups of memory (100 files per group)
Upper and lower preset alarms
Easy-to-read, 4-digit LCD with backlight display
USB connectivity to printer or PC
4 Hz update frequency range
This gauge is designed to measure the thickness of metallic and nonmetallic materials such as steel, aluminum, titanium, plastics, ceramics, glass, and any other good ultrasonic wave conductor, as long as it has parallel top and bottom surfaces. The multifunctional gauge provides basic measurement, scanning, adjustable sound velocity, USB output, and extended storage capabilities for additional flexibility.
The highly innovative gauge design features an intuitive interface that makes setup and operation easy. Simply calibrate the gauge to a known thickness or sound velocity and begin taking measurements. Unit accurately displays measurement results in either inch or millimeter.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Phase II UTG-2800 Ultrasonic Thickness Gauge
Product Type Ultrasonic Thickness Gauge,Display 4-digit LCD,Dimensions " W 3
EW-54103-05
In stock.
$795.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Phase II Digital Ultrasonic Thickness Gauge
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Surface Testing
>
Thickness Gauges
> Phase II Digital Ultrasonic Thickness Gauge
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title