Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Brand:
Product Type:
Price Range:
Class:
Solids (2)
Purity Grade:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 

Phloxine B

Phloxine B
Choose from our comprehensive line of Phloxine B and other Phenyls to Phospho products.
 Product Detail
Sort
 Availability
 Price

Phloxine B, high purity biological stain (25g)

  • Product Type: Organic Chemicals
  • CAS Number: 18472-87-2
  • Form: Solid
EW-88202-43
Usually ships in 2 days.
$142.00 USD/EACH

Qty:

Phloxine B (Certified Biological Stain) (25g)

  • Product Type: Chemical Dyes
  • Purity grade: Certified Biological Stain
  • CAS Number: 18472-87-2
EW-88079-41
Usually ships in 2 days.
$225.00 USD/EACH

Qty:

Phloxine B, high purity biological stain (5g)

  • Product Type: Organic Chemicals
  • CAS Number: 18472-87-2
  • Form: Powder
EW-88202-42
Usually ships in 2 days.
$45.75 USD/EACH

Qty:

3 results (showing 1 - 3 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#