Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Phosphate Buffer Solution, for Chlorine Determination (DPD Methods)

Phosphate Buffer Solution, for Chlorine Determination (DPD Methods)

Phosphate Buffer Solution, for Chlorine Determination (DPD Methods)
Choose from our comprehensive line of Phosphate Buffer Solution, for Chlorine Determination (DPD Methods) and other Chemicals P products.
 Product Detail
Sort
 Availability
 Price

Phosphate Buffer Solution, for Chlorine Determination (DPD Methods) (500 mL)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 6381-92-6, 7487-94-7, 7778-77-0, 7558-79-4 and 7732-18-5
EW-86974-46
Usually ships in 14 days.
$36.50 USD/EACH

Qty:

Phosphate Buffer Solution, for Chlorine Determination (DPD Methods) (1 gal)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 6381-92-6, 7487-94-7, 7778-77-0, 7558-79-4 and 7732-18-5
EW-86986-68
Usually ships in 14 days.
$154.00 USD/EACH

Qty:

Phosphate Buffer Solution, for Chlorine Determination (DPD Methods) (1 L)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 6381-92-6, 7487-94-7, 7778-77-0, 7558-79-4 and 7732-18-5
EW-86981-23
Usually ships in 14 days.
$49.50 USD/EACH

Qty:

3 results (showing 1 - 3 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Phosphate Buffer Solution, for Chlorine Determination (DPD Methods)