Home  >  PicoLite One-Trip Temperature Data Logger
PicoLite One-Trip Temperature Data Logger
PicoLite One-Trip Temperature Data Logger
  • Low-cost, single-use logger is designed specifically for cold chain shipments
  • Unique boomerang feature automatically creates and e-mails a report when logger is connected to a PC
  • High-accuracy internal temperature sensor
  • Direct USB interface for PC communication
  • Software-enabled activation button to start/stop logging and mark time stamps
  • Built-in bi-color LED for logging and immediate alarm indication
  • Six-month battery life at one-minute sampling rate
The compact and easy-to-use PicoLite USB data logger provides a simple single-use solution for temperature monitoring, throughout mass transportation cold chain processes. The PicoLite logger reduces the overhead related to more complex multi-trip data loggers by combining low cost with reliability for single-trip data logging.
Featuring a built-in temperature sensor, activation button, bi-color LED, and direct USB connectivity, this logger offers a "plug and record" solution for every application. The PicoLite USB data logger is supported by free downloadable DataSuite software for comprehensive data analysis, reporting, and alarm notifications (e-mail and SMS notification).
 Product Detail
Sort
 Availability
 Price
EW-18000-07 PicoLite Temperature USB Data Logger
PicoLite Temperature USB Data Logger
Product Type USB Temperature Data Logger,CE Compliance Yes,Connections USB 2.0 compliant
EW-18000-07
Call 1-888-358-4705 for availability.
$21.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  PicoLite One-Trip Temperature Data Logger
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title