log in or register
Material:
Acetal (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Quick-Disconnect Fittings > Plastic Quick-Disconnect Couplings (APC Series) > Plastic Quick Disconnect Fittings, Barbed Body
Plastic Quick Disconnect Fittings, Barbed Body
Plastic Quick Disconnect Fittings, Barbed Body
  • Acetal fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone material meet USP Class VI requirements
Simply press the thumb latch to disconnect coupling halves. Polycarbonate and polysulfone materials meet USP Class VI requirements and pass all physicochemical, cytotoxicity, leachables, and extractables tests. Order complete couplings or separate bodies and inserts from the table below. 10 fittings/pk.*
 Product Detail
Sort
 Availability
 Price
Representative photo only All Plastic Quick Disconnect Hose Barb Coupling Body with Lock PS 3 8 Tube ID 5 pack
All-Plastic Quick Disconnect Hose Barb Coupling Body with Lock; PS, 3/8" Tube ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.375,Material Polysulfone
EW-31311-27
In stock.
$59.00 USD/PACKAGE
Qty:
Representative photo only All Plastic Quick Disconnect Barbed Body PS 1 2 Tube ID 5 pack
All-Plastic Quick Disconnect Barbed Body; PS, 1/2" Tube ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.5,Material Polysulfone
EW-31311-23
In stock.
$53.00 USD/PACKAGE
Qty:
Representative photo only Quick disconnect fittings Polysulfone fittings 3 8 ID body 5 pack
Quick-disconnect fittings, Polysulfone fittings, 3/8" ID, body, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.375,Material Polysulfone
EW-31311-22
In stock.
$25.00 USD/PACKAGE
Qty:
Representative photo only Quick disconnect fittings Polysulfone fittings 1 4 ID body 5 pack
Quick-disconnect fittings, Polysulfone fittings, 1/4" ID, body, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.25,Material Polysulfone
EW-31311-21
In stock.
$25.00 USD/PACKAGE
Qty:
Representative photo only All Plastic Quick Disconnect Hose Barb Coupling Body with Lock PC 1 4 Tube ID 5 pack
All-Plastic Quick Disconnect Hose Barb Coupling Body with Lock; PC, 1/4" Tube ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.25,Material Polycarbonate
EW-31311-16
In stock.
$27.00 USD/PACKAGE
Qty:
Representative photo only All Plastic Quick Disconnect Hose Barb Coupling Body with Lock PC 1 2 Tube ID 5 pack
All-Plastic Quick Disconnect Hose Barb Coupling Body with Lock; PC, 1/2" Tube ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.5,Material Polycarbonate
EW-31311-18
Usually ships in 9 days or sooner.
$60.50 USD/PACKAGE
Qty:
Representative photo only All Plastic Quick Disconnect Hose Barb Coupling Body with Lock PC 3 8 Tube ID 5 pack
All-Plastic Quick Disconnect Hose Barb Coupling Body with Lock; PC, 3/8" Tube ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.375,Material Polycarbonate
EW-31311-17
In stock.
$27.00 USD/PACKAGE
Qty:
Representative photo only Quick disconnect fittings Polycarbonate fittings PP 1 4 ID body 5 pack
Quick-disconnect fittings, Polycarbonate fittings, PP, 1/4" ID, body, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.25,Material Polycarbonate
EW-31311-11
In stock.
$19.50 USD/PACKAGE
Qty:
Representative photo only All Plastic Quick Disconnect Hose Barb Coupling Body with Lock PS 1 2 Tube ID 5 pack
All-Plastic Quick Disconnect Hose Barb Coupling Body with Lock; PS, 1/2" Tube ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.5,Material Polysulfone
EW-31311-28
In stock.
$51.00 USD/PACKAGE
Qty:
Representative photo only All Plastic Quick Disconnect Hose Barb Coupling Body with Lock PS 1 4 Tube ID 5 pack
All-Plastic Quick Disconnect Hose Barb Coupling Body with Lock; PS, 1/4" Tube ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.25,Material Polysulfone
EW-31311-26
In stock.
$57.00 USD/PACKAGE
Qty:
Representative photo only Quick disconnect body acetal 3 8
Quick-disconnect body, acetal, 3/8
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.375,Material Acetal
EW-06360-95
In stock.
$31.00 USD/PACKAGE of 10
Qty:
Representative photo only Quick disconnect fittings Polycarbonate fittings PP 3 8 ID body 5 pack
Quick-disconnect fittings, Polycarbonate fittings, PP, 3/8" ID, body, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.375,Material Polycarbonate
EW-31311-12
In stock.
$19.50 USD/PACKAGE
Qty:
Representative photo only Quick disconnect body acetal 1 4 tubing ID pack of 10
Quick-disconnect body, acetal, 1/4" tubing ID, pack of 10
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.25,Material Acetal
EW-06360-94
In stock.
$32.00 USD/PACKAGE of 10
Qty:
Representative photo only Quick disconnect fittings Polycarbonate fittings body 1 2 ID 5 pack
Quick-disconnect fittings, Polycarbonate fittings, body, 1/2" ID, 5/pack
Product Type Plastic Quick-Disconnects,Tubing ID in. 0.5,Material Polycarbonate
EW-31311-13
In stock.
$20.00 USD/PACKAGE
Qty:
14 results (showing 1 - 14 )
Per Page 
Home  >  Fittings > Quick-Disconnect Fittings > Plastic Quick-Disconnect Couplings (APC Series) > Plastic Quick Disconnect Fittings, Barbed Body
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title