Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Plastic Storage Bins & Containers

Plastic Storage Bins & Containers

Plastic Storage Bins & Containers

A lab carrier and other storage bins are essential to ensure order in the laboratory or workshop. A lab carrier can greatly lessen the number of spills and levels of breakage that occur in the lab. Whether you need a lab carrier for your reagent bottles, syringes, thread fittings or a tote box to transport glassware, these containers and utility carriers make organization easy. A lab carrier is for more than just bottles and glassware. You can buy a lab carrier that can store odd shaped bottles and even test tube racks. For quick benchtop or shelf access, use open storage bins. You can also consider wall-mount panels of bins or stackable bins to conserve valuable bench space. You can also use multi-drawer storage cabinets to find just what you need quickly. Larger and heavier items such as reagent bottles or bulky equipment may require a tote box for storage. Available from 0.5 to 3.7 cubic feet, HDPE tote boxes are stackable to maximize your space. Tote boxes feature handles for easy transport. Optional lids are sold separately. If you need help selecting a lab carrier or any other storage item contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price

Cole-Parmer Economy Bottle Carrier for 3.5" or Less Diameter Bottles

  • Product Type: Carriers
  • Qty/ea: 1
  • Brand: Cole-Parmer
EW-06722-04
In stock.
$31.00 USD/EACH

Qty:

Thermo Scientific Nalgene Utility Carriers, PP

  • Product Type: Carriers
  • Material: Polypropylene (PP)
  • Length: 14.875 in.
EW-06722-02
In stock.
$42.25 USD/EACH

Qty:

Thermo Scientific Nalgene Utility Carrier, HDPE

  • Product Type: Carriers
  • Material: HDPE
  • Length: 14.875 in.
EW-06722-00
In stock.
$38.75 USD/EACH

Qty:

Polystyrene Trans-Boxes container; 1 section, 8" x 4" x 1-3/16", 24/pack

  • Product Type: Storage Bins
  • Dimensions: 8-1/4" x 4-1/2" x 1-3/8" (outer)
    8" x 4" x 1-3/16" (interior)
  • No. of holders: 1
EW-06816-32
Usually ships in 11 days.
$121.00 USD/PACKAGE of 24

Qty:

Storage Cabinet, 15-Drawer; 17" x 11" x 11"

  • Product Type: Storage Cabinets
  • Dimensions: 17 in x 11 in x 11 in
  • Dimensions: 17 " W
EW-09786-03
In stock.
$211.00 USD/EACH

Qty:

Tip-Out Bin System, 4-Bin Capacity; 23-5/8" x 8-1/6" x 6-5/8"

  • Product Type: Storage Bins
  • Dimensions: 23.625 in W x 8.167 in H x 6.625 in D
  • Dimensions: 23.63 " W
EW-47110-06
In stock.
$79.00 USD/EACH

Qty:

Collapsible Mesh PP Storage Basket (without casters); Blue

  • Product Type: Storage Bins
  • Dimensions: 52.7 x 34.9 x 27.3 cm
  • Dimensions: 13-3/4 " W
EW-06818-70
In stock.
$50.25 USD/EACH

Qty:

Tip-Out Bin Unit, 3 bin capacity 23.625 in W x 9.5 in H x 7.75 in D

  • Product Type: Storage Bins
  • Dimensions: 23.625 in W x 9.5 in H x 7.75 in D
  • Dimensions: 23.63 " W
EW-47110-08
In stock.
$83.75 USD/EACH

Qty:

HDPE Storage Tote Bo x (without Lid), 3.7 cu ft, 26.63" x 18.25" x 14.88"

  • Product Type: Storage Bins
  • Dimensions: 26.625 in x 18.25 in x 14.875 in
  • Dimensions: 18.25 " W
EW-06810-90
In stock.
$91.50 USD/EACH

Qty:

HDPE Lid for 06810-90, 3.7 cu ft Storage Tote Box

  • Product Type: Tote Box Lid
  • Material: HDPE
  • Qty/ea: 1
EW-06810-95
In stock.
$40.00 USD/EACH

Qty:

Tip-Out Bin System, 6-Bin Capacity; 23-5/8" x 4-1/2" x 3-5/8"

  • Product Type: Storage Bins
  • Dimensions: 23.625 in W x 4.5 in H x 3.625 in D
  • Dimensions: 23.63 " W
EW-47110-02
Usually ships in 2 days.
$36.50 USD/EACH

Qty:

ESD AkroBins, 8-1/4"W x 7"H x 14-3/4"D Outer Dimensions

  • Product Type: Storage Bins
  • Dimensions: 8.25 in W x 7 in H x 14.75 in D
  • Dimensions: 8.25 " W
EW-47151-42
In stock.
$42.00 USD/EACH

Qty:

K-resin Trans-Boxes container; 1 section, 12-1/2" x 8-1/4" x 2", 12/pack

  • Product Type: Storage Bins
  • Dimensions: 13-1/8" W x 9" H x 2-5/16" D
  • No. of holders: 1
EW-06816-42
Usually ships in 10 days.
$184.00 USD/PACKAGE of 12

Qty:

K-resin Trans-Boxes container; 24 sections, 2" x 2" x 2", 12/pack

  • Product Type: Storage Bins
  • Dimensions: 13-1/8" x 9" x 2-5/16"
  • No. of holders: 24
EW-06816-45
Usually ships in 10 days.
$179.00 USD/PACKAGE of 12

Qty:

Akrobins PP Storage Bin, 5-1/2" x 5" x 10-7/8"; 1/Pk

  • Product Type: Storage Bins
  • Dimensions: 5.5 in x 5 in x 10.875 in
  • Dimensions: 5.5 " W
EW-06811-26
In stock.
$9.25 USD/EACH

Qty:

89 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Plastic Storage Bins & Containers