log in or register
Price Range:
Availability:
Material:
PVDF (3)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Sanitary Fittings, and Clamps > Plastic sanitary clamp to single hose barb adapters
Plastic sanitary clamp to single hose barb adapters
Plastic sanitary clamp to single hose barb adapters
  • Meet USP Class VI and FDA requirements for food contact
Tapered funnel design reduces “dead space” for exceptionally smooth fluid flow. Smooth inner surface prevents particle entrapment; seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth. Sterilize by gamma irradiation. Choose from three polymers; order silicone gasket separately below.
 Product Detail
Sort
 Availability
 Price
Representative photo only Plastic sanitary couplings tubing ID 1 4 PP 1 9 16 x 31 32 x 3 16
Plastic sanitary couplings; tubing ID, 1/4"; PP; 1 9/16" x 31/32" x 3/16"
Product Type Fittings,Tubing ID in. 0.25,Material Polypropylene
EW-31512-00
In stock.
$27.00 USD/EACH
Qty:
EW-31512-02 Sanitary fittings, Plastic sanitary couplings, PP,1/2" ID, 1-13/16, 31/32", 3/8"
Sanitary fittings, Plastic sanitary couplings, PP,1/2" ID, 1-13/16, 31/32", 3/8"
Product Type Fittings,Tubing ID in. 0.5,Material Polypropylene
EW-31512-02
In stock.
$29.00 USD/EACH
Qty:
Representative photo only plastic sanitary couplings tubing ID 3 8 PP 1 11 16 x 31 32 x 5 16
plastic sanitary couplings; tubing ID, 3/8"; PP; 1 11/16" x 31/32" x 5/16"
Product Type Fittings,Tubing ID in. 0.375,Material Polypropylene
EW-31512-01
In stock.
$28.00 USD/EACH
Qty:
Representative photo only Sanitary fittings Plastic sanitary couplings polysulfone 1 4 ID 1 9 16 31 32 3 16
Sanitary fittings, Plastic sanitary couplings, polysulfone, 1/4" ID, 1-9/16", 31/32", 3/16"
Product Type Fittings,Tubing ID in. 0.25,Material Polysulfone
EW-31512-06
In stock.
$43.00 USD/EACH
Qty:
Representative photo only plastic sanitary couplings tubing ID 3 8 PS 1 11 16 x 31 32 x 5 16
plastic sanitary couplings; tubing ID, 3/8"; PS; 1 11/16 x 31/32" x 5/16"
Product Type Fittings,Tubing ID in. 0.375,Material Polysulfone
EW-31512-07
In stock.
$45.00 USD/EACH
Qty:
Representative photo only plastic sanitary couplings tubing ID 3 8 PVDF 1 11 16 x 31 32 x 5 16
plastic sanitary couplings; tubing ID, 3/8"; PVDF; 1 11/16 x 31/32" x 5/16"
Product Type Fittings,Tubing ID in. 0.375,Material PVDF
EW-31512-04
In stock.
$39.00 USD/EACH
Qty:
EW-31512-08 plastic sanitary couplings; tubing ID, 1/2"; PS; 1 13/16 x 31/32" x 3/8"
plastic sanitary couplings; tubing ID, 1/2"; PS; 1 13/16 x 31/32" x 3/8"
Product Type Fittings,Tubing ID in. 0.5,Material Polysulfone
EW-31512-08
In stock.
$46.00 USD/EACH
Qty:
Representative photo only Sanitary fittings Plastic sanitary couplings PVDF 1 4 ID 1 9 16 31 32 3 16
Sanitary fittings, Plastic sanitary couplings, PVDF,1/4" ID, 1-9/16", 31/32", 3/16"
Product Type Fittings,Tubing ID in. 0.25,Material PVDF
EW-31512-03
In stock.
$39.00 USD/EACH
Qty:
EW-31512-05 Sanitary fittings, Plastic sanitary couplings, PVDF,1/2" ID1-13/16, 31/32", 3/8"
Sanitary fittings, Plastic sanitary couplings, PVDF,1/2" ID1-13/16, 31/32", 3/8"
Product Type Fittings,Tubing ID in. 0.5,Material PVDF
EW-31512-05
Usually ships in 14 days.
$40.00 USD/EACH
Qty:
9 results (showing 1 - 9 )
Per Page 
Home  >  Fittings > Sanitary Fittings, and Clamps > Plastic sanitary clamp to single hose barb adapters
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title