United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Safety Products
>
Gas Detectors
>
Single Gas Detection
> Pocket Ozone Detector
Pocket Ozone Detector
Accurate, easy-to-read digital display
Versatile meter can be used for fixed or portable monitoring
Ideal for leak checks in generators, valves and piping
The heated metal oxide semiconductor has a range of 0.02 to 10 ppm; digital readout is in 0.01 ppm increments. Use as a permanent monitor indoors when the concentration average is below 0.1 ppm; use outdoors or at concentrations up to 10 ppm for short periods of time. Includes rechargeable batteries, charger, and carrying case.
Order optional calibrator to verify that your ozone monitor is conforming to the original calibration. Calibrator recalibrates detector at 0.1 ppm. It measures 21⁄4''W x 33⁄4''H x 1''D and includes a 9 V battery.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Digital ozone monitor
Product Type Single Gas Monitors/Detectors,Model A-21ZX
EW-86316-30
Call 1-888-358-4705 for availability.
$750.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Pocket Ozone Detector
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Safety Products
>
Gas Detectors
>
Single Gas Detection
> Pocket Ozone Detector
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title