United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Availability:
In Stock
(1)
CE Compliance:
Yes
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Baths and Circulators
>
Immersion Circulators
> PolyScience Immersion Circulators
PolyScience Immersion Circulators
Converts any tank into a user-friendly circulating bath
Low level cutoff with proportional temperature control
User-settable high temperature cutoff
All wetted parts are 304 and 316 stainless steel
These immersion circulators features proportional temperature control and high-temperature and low-level cutoff. Advanced multi-speed pump minimizes turbulence in small tanks while providing greater uniformity in large tanks. Directable nozzle lets you set the direction of pump flow and accepts 13-mm ID tubing.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
PolyScience Standard Immersion Circulator, 12L, 120 VAC
Product Type Circulators (Immersion),Capacity Liters 12,Min temperature º C Ambient
EW-12101-84
In stock.
$1,714.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
PolyScience Immersion Circulators
PolyScience Standard Immersion Circulator, 12L, 240 VAC
Product Type Circulators (Immersion),Capacity Liters 12,Min temperature º C Ambient
EW-12101-86
Usually ships in 14 days.
$1,702.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
PolyScience Immersion Circulators
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Baths and Circulators
>
Immersion Circulators
> PolyScience Immersion Circulators
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title