United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Heating Equipment
>
Heating Torches
> Portable Torch-Burner
Portable Torch-Burner
Use as both a handheld torch and portable burner
Butane fuel is self-contained and can be refilled with standard butane canister fuel
Single-knob gas control with trigger-activated electronic auto ignition
One fuel canister provides over 2500°F (1371°C) and up to 1.5 hours of continuous burn time
This unit can be used as both a torch and a burner. The torch can be inserted in the provided black base or turned on its side with nozzle pointing directly upward resting on the included metal ring
stand to simulate a benchtop burner.
This affordable torch burner contains a completely controlled flame size that can be manually controlled for off/on or set for a continuous flame. Because the unit will not flare in any position,
this torch can be used as a portable burner when used with included metal stand. There is an ON/OFF switch for safety. Compact size (5 1⁄2''(14 cm) L x 3 1⁄2''(9 cm) W x 8''(21 cm) H) make this an excellent burner for field use.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Portable torch burner
Product Type Accessory
EW-36310-41
In stock.
$83.75 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Portable Torch-Burner
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Heating Equipment
>
Heating Torches
> Portable Torch-Burner
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title