Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Brand:
Product Type:
Availability:
Form:
Liquid (3)
Quantity (g):
500 (2)
2500 (1)
125 (1)
Quantity (mL):
500 (1)
1000 (1)
120 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Potassium Iodide

Potassium Iodide
Choose from our comprehensive line of Potassium Iodide and other Potassium products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Potassium Iodide Granular Acs 500g

Potassium Iodide Granular Acs 500g

Product Type Chemical Reagents,Purity grade ACS,Form Crystal
EW-89100-47
Usually ships in 5 days.
$305.00 USD/EACH

Qty:

Representative photo only Potassium Iodide Granular Usp 2 5kg

Potassium Iodide Granular Usp 2.5kg

Product Type Chemical Reagents,Purity grade USP,Form Granular
EW-89100-50
Usually ships in 9 days.
$1,450.00 USD/EACH

Qty:

Representative photo only Potassium Iodide Granular Acs 125g

Potassium Iodide Granular Acs 125g

Product Type Chemical Reagents,Purity grade ACS,Form Crystal
EW-89100-48
Usually ships in 4 days.
$94.00 USD/EACH

Qty:

Representative photo only Potassium Iodide Granular Free Flowing Certified ACS 500g

Potassium Iodide (Granular, Free Flowing/Certified ACS) (500g)

Product Type Chemical Reagents,Purity grade Certified ACS,Form Granular
EW-88020-56
In stock.
$610.00 USD/EACH

Qty:

Representative photo only Potassium Iodide Granular Usp 500g

Potassium Iodide Granular Usp 500g

Product Type Chemical Reagents,Purity grade USP,Form Granular
EW-89100-49
Usually ships in 6 days.
$210.00 USD/EACH

Qty:

Representative photo only Potassium iodide reagent ACS granular 500g

Potassium iodide, reagent ACS granular (500g)

Product Type Chemical Reagents,Form Granular
EW-88355-47
In stock.
$193.00 USD/EACH

Qty:

Representative photo only Potassium Iodide 10 W v 1L

Potassium Iodide 10% W/v 1L

Product Type Chemical Reagents,Form Liquid,Quantity mL 1000
EW-88085-04
In stock.
$92.00 USD/EACH

Qty:

Representative photo only Potassium Iodide 0 5 w v Aqueous Solution 120 mL

Potassium Iodide, 0.5% (w/v) Aqueous Solution (120 mL)

Product Type Chemical Reagents,Form Liquid,Quantity mL 120
EW-86991-89
Usually ships in 10 days.
$20.00 USD/EACH

Qty:

Representative photo only Potassium Iodide 0 5 w v Aqueous Solution 500 mL

Potassium Iodide, 0.5% (w/v) Aqueous Solution (500 mL)

Product Type Chemical Reagents,Form Liquid,Quantity mL 500
EW-86975-08
Usually ships in 13 days.
$39.00 USD/EACH

Qty:

Representative photo only Potassium iodide briquettes 99 2 5kg

Potassium iodide, briquettes 99% (2.5kg)

Product Type Chemical Reagents
EW-88210-02
In stock.
$590.00 USD/EACH

Qty:

Representative photo only Potassium iodide briquettes 99 500g

Potassium iodide, briquettes 99% (500g)

Product Type Chemical Reagents
EW-88210-03
In stock.
$153.00 USD/EACH

Qty:

Representative photo only Potassium Iodide Cert Acs 10kg

Potassium Iodide Cert Acs 10kg

Product Type Chemical Reagents
EW-88079-44
In stock.
$2,432.00 USD/EACH

Qty:

Representative photo only Potassium iodide 99 995 50g

Potassium iodide 99.995% (50g)

Product Type Chemical Reagents
EW-88206-47
In stock.
$250.00 USD/EACH

Qty:

Representative photo only Potassium Iodide White Free Flowing Granules 500g

Potassium Iodide (White, Free-Flowing Granules) (500g)

Product Type Chemical Bio-Reagents,Form Granular,Hazard symbol Irritant
EW-88066-94
In stock.
$303.00 USD/EACH

Qty:

Representative photo only Potassium Iodide Granular Free Flowing USP FCC 3kg

Potassium Iodide (Granular, Free Flowing/USP/FCC) (3kg)

Product Type Chemical Reagents,Purity grade USP/FCC,Form Granular
EW-88020-62
In stock.
$1,202.00 USD/EACH

Qty:

19 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title