Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Material Testing & Physical Testing
>
Surface Testing
>
Surface Roughness Testers
> Precision Surface Roughness Tester
Precision Surface Roughness Tester
Provides precision accurate surface profile testing quickly and easily
This state-of-the-art surface roughness tester is designed to instill the highest level of confidence in your production, shop floor, or QC lab roughness testing measurements. Unit provides fifteen different popular surface roughness scales including Ra, Rq (Rms), Rt, Rz, and Rmax, and features extended memory and RS-232/USB output, making it ideal for most surface roughness testing applications.
The hypersensitive, sophisticated diamond stylus provides 0.001 (µm) micron display resolution. Set up and operation is simple and unit allows you to easily make changes for in/mm (µin/µm), cut-off length, sampling length, parameters, level of test (stylus position), digital filters, and more. The portable, battery powered capability provides additional flexibility and the large LCD display makes operator interpretation of measurements easy.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Phase-II SRG-4500 Portable Surface Roughness Tester
Dimensions " W 2-3/4,Dimensions " H 6,Dimensions " D 2-1/2
EW-54103-06
Usually ships in 4 days.
$1,840.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Precision Surface Roughness Tester
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Material Testing & Physical Testing
>
Surface Testing
>
Surface Roughness Testers
> Precision Surface Roughness Tester
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity