United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pumps
>
Metering Pumps
>
Piston Pumps
> Preparative Digital Gradient Piston Pump
Preparative Digital Gradient Piston Pump
Deliver precise volumes at pressures up to 6000 psi
Ideal for use in HPLC applications
Pulse dampener significantly reduces pulsation with low dead volume.
Preparative digital pump provides high flow rates and high pressure capabilities with consistent performance. Dual ball and seat check valves in each head reduce pulsation.
Microprocessor controls motor drive circuits, interfaces with the chemical-resistant keypad and digital LED display, receives signals to refill based on flow rate, and provides RS-232 communication. Pressure control feature provides real-time pressure monitoring with upper and lower pressure limits. Other features include prime/purge valves for easy solvent change and priming capabilities, auto-flush piston wash, run/stop inputs, and dual-voltage capabilities.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Preparative Digital High-Pressure Gradient Piston Pump
Product Type Piston Metering Pumps,Max pressure 4000 psi,Min flow rate mL/min 0.1
EW-74930-20
Usually ships in 13 days.
$7,455.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Preparative Digital Gradient Piston Pump
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Pumps
>
Metering Pumps
>
Piston Pumps
> Preparative Digital Gradient Piston Pump
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title