Product Type Agar,Description PDA (Potato Dextrose Agar), for cultivation of yeasts and mold and for the inducement of sporulation,Qty/pk 10
Usually ships in 7 days.
Product Type Agar,Description TSA, general growth medium for the isolation and cultivation of microorganisms,Qty/pk 10
Usually ships in 14 days.
Product Type Agar,Description Blood Agar, 5%, general purpose growth media for the isolation, cultivation, and differentiation of a wife variety of microorgansim,Qty/pk 10
Usually ships in 4 days.
Product Type Agar,Description Sab Dex, for the isolation, cultivation, and maintenance of nonpathogenic and for pathogenic species of fungi and yeasts,Qty/pk 10
Usually ships in 7 days.
Product Type Agar,Description Standard Methods Agar, for determining the microbial content in dairy products, food, water and other material of sanitary importance,Qty/pk 10
Usually ships in 5 days.
Product Type Agar,Description Mannitol Salt Agar, selective and differential medium for the isolation of pathogenic staphylococci,Qty/pk 10
Usually ships in 4 days.
Product Type Agar,Description Cetrimide Agar, recommended for the selective isolation and identification of Pseudomonas aeruginosa,Qty/pk 10
Usually ships in 6 days.
Product Type Agar,Description MacConkey Agar with Sorbital, selective and differential medium for the detection of enterohemorrhagic Escherichia Coli 0157:H7,Qty/pk 10
Usually ships in 5 days.
Product Type Agar,Description Baird Parker Agar, selective medium for the detection and enumeration of coagulase-positive Staphylococci from food samples,Qty/pk 10
Usually ships in 5 days.
Product Type Agar,Description HE Enteric Agar, selective and differential medium for the isolation and differentiation of gram-negative enteric pathogens,Qty/pk 10
Usually ships in 4 days.
Product Type Agar,Description XLD Agar, selective and differential medium for the isolation of gram-negative enteric pathogens from fecal and other specimen,Qty/pk 10
Usually ships in 4 days.